Skip to Content
Merck
All Photos(1)

Key Documents

05495

Sigma-Aldrich

Silver proteinate

~8% Ag basis

Synonym(s):

Albumose silver, Protargol, Silver protein, strong

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12171500
NACRES:
NA.47

concentration

~8% Ag

pH

8.0-9.5 (10 mg/mL in H2O)

Looking for similar products? Visit Product Comparison Guide

Application

Silver proteinate (Protargol) is used as a component of the Periodic Acid-ThioSemiCarbazide-Silver Proteinate (PA-TSC-SP) staining protocol for ultrastructural detection of carbohydrates such as glycogen, and proteoglycans.

Other Notes

Protargol silver impregnation and neuronal selectivity - light and electron microscopic observations

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bettina Sonntag et al.
The Journal of eukaryotic microbiology, 51(6), 670-677 (2005-01-26)
Two euplanktonic ciliates, Urotricha psenneri n. sp. (Prostomatida) and Amphileptus piger (Vuxanovici, 1962) n. comb. (Pleurostomatida), were discovered in the surface plankton of the oligotrophic Lake Traunsee in Austria. Their morphology and infraciliature were studied in live cells as well
Hongwei Ma et al.
The Journal of eukaryotic microbiology, 53(2), 85-95 (2006-04-04)
Two epibiotic peritrichs infested the blue crab, Callinectes sapidus, from the Gulf of Mexico, Mississippi, USA. Epistylis callinectes n. sp. was isolated from the epipods of maxillipeds, bases of gill-cleaning setae, and gills, and Epistylis clampi n. sp. was isolated
Xin Gao et al.
European journal of protistology, 47(3), 235-237 (2011-04-19)
During conjugation of Paramecium caudatum, there are two well-known stages when nuclear migration occurs. What happens to the nuclei is closely related to their localisations in cells. The first of these stages is the entrance of one meiotic product into
V S Minasian et al.
Vestnik otorinolaringologii, (6)(6), 36-37 (2005-02-09)
The authors review current views on the treatment of acute rhinitis (AR) in infants and present their experience in AR treatment of 30 infants aged from 3 days to 1 year. Combined treatment with Physiomer baby quickly eradicated inflammation. The
Renu Gupta et al.
European journal of protistology, 42(1), 29-48 (2006-10-31)
The oxytrichid ciliate Architricha indica nov. gen., nov. sp., isolated from the river Yamuna, Delhi, shows a new combination of characters. It possesses a flexible body, 18 frontal-ventral-transverse (FVT) cirri, 3 right and 2 left marginal cirral rows, 6 dorsal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service