Stericup® Quick Release, or “QR”, filters feature a connection between funnel filter and receiver bottle that requires only a quarter-turn release to facilitate funnel removal. These filters come fully engaged – there is no need to tighten before use. The receiver bottle cap has flanges on the top surface to improve grip, as well as a positive lock stop to ensure a leak-proof seal with the receiver bottle. Other improvements include a large, frosted writing surface on the receiver bottle and a lighter cap color to facilitate labelling, more prominent volume markings on the filtration funnel and receiver bottle, and a product label with a peel-off lot record strip on the bag for easy viewing in storage.
Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
Quality Segment
material
polyethersulfone membrane, polystyrene bottle, polystyrene funnel(s), polystyrene housing
product line
Stericup®
feature
hydrophilic
packaging
pack of 12 ea
parameter
45 °C max. temp.
description
2.4 mL (hold-up volume)
technique(s)
sterile filtration: suitable
H
120 mm
diam.
12.5 cm
filter capacity
150 mL
filter diam.
73 mm
filtration area
40 cm2
matrix
Express PLUS™
pore size
0.22 μm pore size
fitting
inlet funnel, outlet flask (Receiver)
shipped in
ambient
storage temp.
no temp limit
General description
Stericup® and Steritop® sterile filtration devices use reliable, high-performance membranes for optimum sterile filtration of laboratory reagents, buffers, and media. These devices combine superior flow rates and throughput with low non-specific binding and a stable, no-tip design.
This Stericup® 0.22 μm pore size polyethersulfone (PES) membrane filter unit combines a 150 mL bottle-top with a 150 mL receiver flask and cap for processing and storage.
Application
- Tissue culture media +/- additives
- Buffers
- Biological solutions
Features and Benefits
- The Quick Release filter funnel disconnects from the receiver bottle with just a quarter turn—reducing the likelihood of spillage and making it easier to manipulate on the bench or under the hood.
- Bold product identification on the Steritop® filter funnel plainly displays membrane characteristics, including composition and pore size, for rapid, application-appropriate product selection.
- Click seal cap is flanged and modified to enable a firm grip in wet or dry conditions—and a tactile stop confirms secure closure. Worry less about the possibility of spilling or contamination of sterilized contents.
- Frosted, ample writing surface on the receiver bottle and lighter cap color facilitate clear labelling to improve legibility and reduce unreadable smears.
- Click seal cap rests on the side to avoid risk of contamination.
- Receiver bottles are stackable to save storage space.
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| description pore size 0.22 μm, polyethersulfone membrane, filter capacity 150 mL, pack of 12 ea | description pore size 0.22 μm, polyethersulfone membrane, filter capacity 1000 mL, pack of 12 ea | description pore size 0.22 μm, polyethersulfone membrane, filter capacity 500 mL, pack of 12 ea | description pore size 0.22 μm, polyethersulfone membrane, filter capacity 250 mL, pack of 12 ea |
| filter capacity 150 mL | filter capacity 1000 mL | filter capacity 500 mL | filter capacity 250 mL |
| pore size 0.22 μm pore size | pore size 0.22 μm pore size | pore size 0.22 μm pore size | pore size 0.22 μm pore size |
| material polystyrene funnel(s), polystyrene housing, polyethersulfone membrane, polystyrene bottle | material polystyrene housing, polystyrene funnel(s), polystyrene bottle, polyethersulfone membrane | material polystyrene bottle, polyethersulfone membrane, polystyrene funnel(s), polystyrene housing | material polystyrene funnel(s), polystyrene housing, polystyrene bottle, polyethersulfone membrane |
| feature hydrophilic | feature hydrophilic | feature hydrophilic | feature hydrophilic |
| packaging pack of 12 ea | packaging pack of 12 ea | packaging pack of 12 ea | packaging pack of 12 ea |
| fitting inlet funnel, outlet flask (Receiver) | fitting outlet flask (Receiver), inlet funnel | fitting outlet flask (Receiver), inlet funnel | fitting inlet funnel, outlet flask (Receiver) |
Still not finding the right product?
Explore all of our products under Millipore® Stericup® Quick Release Vacuum Filtration System
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
What is the difference between classic Stericup® filters and Stericup® “Quick Release” filters?
1 answer-
Helpful?
-
-
Can Stericup® and Steritop® filters be reused?
1 answer-
All Stericup® and Steritop® filters were developed for single use, disposable processing.
Helpful?
-
-
Can we filter oil-type liquids with Stericup® filters?
Can we filter oil-type liquids with Stericup® filters?
1 answer-
Likely not, due to pore size/possible fouling/chemical compatibility. Refer to our guide, "Chemical compatibility of filter components": https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/302/855/membrane-chemical-compatibility-ds2532en-ms.pdf
Helpful?
-
-
What is the maximum vacuum pressure for filtration?
1 answer-
700 mm Hg (27.5” Hg, 0.9 bar) differential vacuum at 25 °C (77 °F)
Helpful?
-
-
What is the maximum processing volume for Stericup® and Steritop® filters? Can I add more volume after initial filtration?
1 answer-
All Stericup® and Steritop® filters were developed for single-use application. The maximum volume is dependent on the receiver bottle capacity. Stericup® filters come with a receiver bottle with a specified maximum volume. For Steritop® filters, the maximum recommended volume is 2 L with an appropriate-sized receiver bottle. Larger processing volumes can result in membrane fouling, increase chance of spillage and contamination, and lead to possible airlock*.
*Note that devices with PES membranes can airlock if filtered to dryness. Some liquid must be left on top of the filter, then the system stopped, receiver changed or emptied, and more sample volume added before restart. This increases the chance of spillage.Helpful?
-
-
Are Stericup® and Steritop® filters GMP-certified?
1 answer-
No, Stericup® and Steritop® devices are not GMP-approved.
Helpful?
-
-
What tubing can be used with Stericup® and Steritop® vacuum filters?
What tubing can be used with Stericup® and Steritop® vacuum filters?
1 answer-
Stericup® and Steritop® filters are fitted with a 1/4” tube adapter. We recommend 3/16 in. (4.8 mm) ID silicone tubing to fit this tube adapter: https://www.sigmaaldrich.com/product/mm/xx7100004
Helpful?
-
-
What leachables are detected during filtration processes using Stericup® filters?
1 answer-
We have not performed extractables/leachables studies for our Stericup® filters.
Helpful?
-
-
The User Guide says “Use only glass or plastic bottles designed for vacuum applications”. Can you specify which bottles are compatible?
1 answer-
Steritop® Quick Release bottle top units require a vacuum-rated receiver bottle with either a GL33 or GL45 mm closure thread as specified. Steritop® E bottle top units require a vacuum-rated receiver bottle with a GL45 mm closure thread. Bottles no larger than 2 L should be used.
Glass bottles, GL33:
• Wheaton media bottles (glass), 500 mL (Product Z364886)
• Wheaton media bottles (glass), 1 L (Product Z364894)
Glass bottles, GL45
• Corning® glass media storage bottle, 125 mL (Product CLS1399125)
• Corning® glass media storage bottle, 250 mL (Product CLS1399250)
• Corning® glass media storage bottle, 500 mL (Product CLS1399500)
• Corning® glass media storage bottle, 1 L (Product CLS13991L)
• Pyrex® round media storage bottle, 500 mL (Product CLS1395500)
• Pyrex® round media storage bottle, 1 L (Product CLS13951L)
• KIMBLE® KIMAX® Media bottle, 250 mL (Product DWK14395250)
• KIMBLE® KIMAX® Media bottle, 500 mL (Product DWK14395500)
• KIMBLE® KIMAX® Media bottle, 1 L (Product DWK143951000)
Plastic (pre-sterile) bottles, GL45:
• Stericup® Quick Release Receiver flask, 250 mL (Product S200B02RE)
• Stericup® Quick Release Receiver flask, 500 mL (Product S200B05RE)
• Stericup® Quick Release Receiver flask, 1000 mL (Product S200B10RE)Helpful?
-
-
Can Stericup® and Steritop® filters be used to prepare solutions that will be administered to patients?
1 answer-
No, all Stericup® and Steritop® devices are Research Use Only.
Helpful?
-



