Skip to Content
Merck

Skip To

MB000000E

m-Endo Total Coliform Broth (Dehydrated)

pH 7.2±0.1 (25.0 °C), powder (dehydrated), for coliforms, pkg of 110 g, suitable for microbiology

Synonym(s):

MF Endo Broth, MF Endo Medium, m-Coliform Broth

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41106213
NACRES:
NA.74
eCl@ss:
32160801
Ph:
7.2±0.1 (25.0 °C)
Form:
powder (dehydrated)
Suitability:
coliforms
Application(s):
clinical
cosmetics
environmental
food and beverages
industrial qc
water monitoring, microbiology
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

description

For testing drinking water for total coliform detection

form

powder (dehydrated)

shelf life

60 mo.

packaging

pkg of 110 g

color

pink-purple dehydrated powder, pink-red prepared medium

input

sample type wine
sample type cosmetics
sample type soft drink
sample type bottled water
sample type beer
sample type cider
sample type sport drink
sample type food(s)
sample type beverage(s)

pH

7.2±0.1 (25.0 °C)

application(s)

clinical
cosmetics
environmental
food and beverages
industrial qc
water monitoring, microbiology

compatibility

for use with MF-method, for use with Microfil®, for use with MicropreSure®, for use with Milliflex®

shipped in

ambient

storage temp.

2-30°C

suitability

coliforms

General description

m-Endo Total Coliform Broth is a dehydrated, selective, and differential medium designed for the detection and enumeration of total coliform bacteria in water samples. The medium is formulated with sodium sulfite and basic fuchsin, which inhibits the growth of Gram-positive bacteria while allowing coliform bacteria to grow. Lactose-fermenting bacteria produce acetaldehyde, which reacts with sodium sulfite and fuchsin to form red colonies. Rapid lactose fermentation results in a metallic sheen on the colonies. Non-lactose fermenting bacteria form colorless colonies.

Application

Type of Test
Bioburden

Microbial Limits
m-Endo Total Coliform Broth is used for testing drinking water to detect total coliforms using membrane filtration techniques. It is suitable for bioburden testing, microbial limits testing, and water monitoring in clinical, environmental, food and beverage, cosmetics, and industrial quality control applications. This media is ideal for laboratories conducting water quality testing and microbial analysis in compliance with standard methods for the examination of water and wastewater.

Features and Benefits

Incubation Temperature: 35 °C ± 0.5
Incubation Time: 24 hrs ± 2
Standard Methods for the Examination of Water and Wastewater

Packaging

Dehydrated 110 g in plastic bottle

Preparation Note

m-Endo

Analysis Note

Target Microorganisms: Total coliforms
Coliform colonies appear deep red with distinct green metallic sheen

Legal Information

MICROFIL is a registered trademark of Merck KGaA, Darmstadt, Germany
MICROPRESURE is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MB000000FMHA000P2EMB000000T
form

powder (dehydrated)

form

powder (dehydrated)

form

liquid

form

powder (dehydrated)

application(s)

clinical
cosmetics
environmental
food and beverages
industrial qc
water monitoring, microbiology

application(s)

clinical
cosmetics
environmental
food and beverages
industrial qc (microbial monitoring)
microbiology
water monitoring

application(s)

bioburden testing
clinical
cosmetics
environmental
food and beverages
industrial qc
microbiology
water monitoring

application(s)

clinical
cosmetics
environmental
food and beverages
industrial qc
water monitoring, microbiology

suitability

coliforms

suitability

fecal coliforms

suitability

Escherichia spp., coliforms

suitability

Aspergillus niger, Bacillus subtilis, Candida albicans, Clostridium sporogenes, Pseudomonas aeruginosa, Staphylococcus auerus, heterotrophic bacteria

storage temp.

2-30°C

storage temp.

2-30°C

storage temp.

2-8°C

storage temp.

2-30°C

shelf life

60 mo.

shelf life

48 mo.

shelf life

12 mo.

shelf life

60 mo.

shipped in

ambient

shipped in

ambient

shipped in

wet ice

shipped in

ambient


pictograms

Health hazard

signalword

Warning

hcodes

Hazard Classifications

Carc. 2

Storage Class

11 - Combustible Solids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters