Skip to Content
Merck

MB000000F

m-FC Broth (Dehydrated) without Rosolic Acid

For testing waste and effluent waters for fecal coliforms. To be used with rosolic acid (MB00 000 0R).

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41106213
NACRES:
NB.24
eCl@ss:
32160801
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

powder (dehydrated)

shelf life

48 mo.

packaging

pkg of 110 g

color

beige dehydrated powder, with slight blue tint, blue prepared medium w/rosolic acid, cranberry prepared medium w/o rosolic acid

input

sample type cosmetics
sample type soft drink
sample type beer
sample type sport drink
sample type wine
sample type food(s)
sample type beverage(s)
sample type bottled water
sample type cider

pH

7.4±0.2 (25.0 °C)

application(s)

clinical
cosmetics
environmental
food and beverages
industrial qc (microbial monitoring)
microbiology
water monitoring

compatibility

for use with MF-method, for use with Microfil®, for use with MicropreSure®, for use with Milliflex®

shipped in

ambient

storage temp.

2-30°C

suitability

fecal coliforms

Application

Type of Test
Bioburden

Microbial Limits

Features and Benefits

Incubation Temperature: 44.5 °C ± 0.2
Incubation Time: 24 hrs ± 2
Standard Methods for the Examination of Water and Wastewater

Packaging

Dehydrated 110 g in plastic bottle

Preparation Note

Rosolic Acid

Analysis Note

Fecal coliforms appear blue, other colonies appear gray to cream colored
Target Microorganisms: Fecal coliforms

Legal Information

MICROFIL is a registered trademark of Merck KGaA, Darmstadt, Germany
MICROPRESURE is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

11 - Combustible Solids

wgk

WGK 3



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library