Skip to Content
Merck
All Photos(2)

Key Documents

MAB2300X

Sigma-Aldrich

Milli-Mark® FluoroPan Neuronal Marker - Alexa488 conjugated

Milli-Mark FluoroPan Neuronal Marker -Alexa488 conjugated is an antibody targeting the FluoroPan Neuronal Marker protein, validated for use in ICC & IF.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 488

clone

monoclonal

species reactivity

mouse, rat, human

manufacturer/tradename

Neuro-Chrom

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable

isotype

IgG1

shipped in

wet ice

target post-translational modification

unmodified

General description

Antibodies to neuronal proteins have become critical tools for identifying neurons and discerning morphological characteristics in culture and complex tissue. While the labeling from classic histological techniques, such as Golgi staining, and modern molecular approaches, such as GFP constructs, yield excellent cytoarchitectural detail, these approaches are technically challenging and impractical for many neuroscience research needs. Neuron-specific antibodies are convenient precision tools useful in revealing cytoarchitecture, but are limited to the protein target distribution within the neuron, which may differ greatly from nucleus to soma to dendrite and axon. To achieve as complete a morphological staining as possible across all parts of neurons, Millipore previously developed a monoclonal antibody blend (Catalog No. MAB2300) that reacts against key somatic, nuclear, dendritic, spine, and axonal proteins distributed across the pan-neuronal architecture. Now Millipore has formulated MAB2300X, an Alexa 488 conjugated version of the pan-neuronal antibody blend MAB2300 to facilitate immunofluorescence studies. Each antibody within the blend is conjugated to Alexa 488 therefore enabling detection of an entire neuron with only one microscopic emission channel. MAB2300X has been validated in diverse fixations, cell culture, and immunohistochemistry protocols, giving researchers a convenient and specific qualitative and quantitative tool for studying neuronal morphology. High morphological resolution has been achieved using MAB2300x including distral dentrites and spines.

Specificity

MAB2300X is specific to axons (neurites), dendrites, spines, nucleus, and the cell body of neurons.
Reactivity with other species has not been determined.

Immunogen

Epitope: Whole Neuron Marker

Application

Immunocytochemistry:

1. Culture neurons as desired. When ready, gently wash with PBS and fix cells with 4% paraformaldehyde in PBS, 30min at room temp (RT).
2. Wash with PBS, 3 x 5min each, at RT and block with blocking buffer (1% BSA, 5% Serum, 0.2% Triton X in PBS) for 1 hr at RT.
3. Incubate cells with Milli-Mark FluoroPan Neuronal Marker (Mouse IgG conjugated with Alexa 488) for 2hr at RT (dilute antibody in blocking buffer).
4. Wash with PBS, 3 x 5min each, and cover slip with LIGHT DIAGNOSTICS Mounting Fluid (Catalog No. #5013). View using fluorescent microscope with FITC filter.
Research Category
Neuroscience
Research Sub Category
Neuronal & Glial Markers

Quality

Routinely tested on cortical neurons in Immunocytochemistry 1:150 - 1:100.



Physical form

Blended monoclonal antibody cocktail in PBS with 0.05% NaN3

Storage and Stability

Maintain at 2-8°C for up to 6 months from date of receipt.

Analysis Note

Control
Cortical neurons

Legal Information

ALEXA FLUOR is a trademark of Life Technologies
MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

IGF-1 enhances cell proliferation and survival during early differentiation of mesenchymal stem cells to neural progenitor-like cells.
Huat, TJ; Khan, AA; Pati, S; Mustafa, Z; Abdullah, JM; Jaafar, H
BMC Neuroscience null
Hong Fang et al.
Molecular medicine reports, 17(1), 1083-1092 (2017-11-09)
General anesthesia is widely used in pediatric surgery, although the influence of general anesthesia on cerebellar information transmission and motor function is unclear. In the present study, neonatal mice received repeated inhalation of sevoflurane, and electrophysiological alterations in Purkinje cells
Clark W Bird et al.
Alcohol (Fayetteville, N.Y.), 86, 17-24 (2020-04-01)
A relatively common polymorphism in the human brain-derived neurotrophic factor (BDNF) gene (Val66Met, which corresponds to Val68Met in mice) has been shown to modulate cognitive function and vulnerability to mental health disorders. This substitution impairs trafficking and activity-dependent release of
Lori A Newman et al.
PloS one, 6(12), e28427-e28427 (2011-12-20)
When administered either systemically or centrally, glucose is a potent enhancer of memory processes. Measures of glucose levels in extracellular fluid in the rat hippocampus during memory tests reveal that these levels are dynamic, decreasing in response to memory tasks
Patricia Rodriguez-Rodriguez et al.
Brain : a journal of neurology, 140(12), 3269-3285 (2017-10-21)
Insulin signalling deficiencies and insulin resistance have been directly linked to the progression of neurodegenerative disorders like Alzheimer's disease. However, to date little is known about the underlying molecular mechanisms or insulin state and distribution in the brain under pathological

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service