Skip to Content
Merck

Skip To

A1595

Bovine Serum Albumin solution

10% in DPBS, low endotoxin, fatty acid free, suitable for cell culture, sterile-filtered

Synonym(s):

Albumin solution from bovine serum, BSA solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.27
MDL number:
eCl@ss:
32160409
Biological source:
bovine
Assay:
90-120 mg/mL protein basis (biuret)
Form:
solution
Technique(s):
cell culture | mammalian: suitable
Impurities:
≤0.05% fatty acid, ≤200 EU/mL endotoxin
Concentration:
10% in DPBS
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

bovine

Quality Segment

sterility

sterile-filtered

product line

BioReagent

assay

90-120 mg/mL protein basis (biuret)

form

solution

purified by

heat shock fractionation

contains

does not contain stabilizer or preservative

origin

USA origin

concentration

10% in DPBS

technique(s)

cell culture | mammalian: suitable

impurities

≤0.05% fatty acid, ≤200 EU/mL endotoxin

pH

7.1-7.7

UniProt accession no.

storage temp.

2-8°C

Gene Information

bovine ... ALB(280717)

Application

Recommended for use as a base for the preparation of defined, fatty acid supplements. Can be loaded with up to 4 moles of fatty acids per mole of albumin.

BSA is a known fatty acid (FA) carrier. Thus in FA-related applications like cell culture, the specific BSA can be important, as related to control over specific fatty acids used to culture cell lines, because different cell lines can be variably sensitive to particular FA;s. Fatty acid-free BSA therefore is useful in cell culture work where the specific fatty acid content must be carefully controlled, so that researchers can control the specific FA′s needed for their particular cell lines. Fatty acid-free BSA also allows for optimal and maximum binding sites for the specific fatty acids that researchers wish to use in their particular cell culture experiments. Use of fatty-acid-free BSA also addresses concerns about endogenous FA′s potentially in non-fatty acid-free BSA.

Features and Benefits

  • Fatty acid free / tested to be low in fatty acid content
  • Low endotoxin
  • Sterile-filtered

Preparation Note

Prepared with Heat Shock Fractionated fatty acid-free albumin in DPBS D5652.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A0336A4919A7030
description

10% in DPBS, low endotoxin, fatty acid free, suitable for cell culture, sterile-filtered

description

30% in saline, IgG free, aseptically filled

description

heat shock fraction, protease free, low endotoxin, suitable for cell culture, pH 7, ≥98%

description

heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%

assay

90-120 mg/mL protein basis (biuret)

assay

28-32% protein basis (biuret)

assay

≥98%

assay

≥98%

biological source

bovine

biological source

bovine

biological source

bovine

biological source

bovine

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

ELISA: suitable, ligand binding assay: suitable, blood typing: suitable

origin

USA origin

origin

USA origin

origin

USA origin

origin

USA origin

Quality Level

300

Quality Level

200

Quality Level

200

Quality Level

300

form

solution

form

solution

form

lyophilized powder

form

lyophilized powder


Still not finding the right product?

Explore all of our products under Bovine Serum Albumin solution


Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. Can I store undiluted aliquots of this solution at -80º or -20º C for longterm storage? If not, how long does this solution last at 2-8ºC?

    1 answer
    1. Undiluted aliquots of this product can be stored at -80º or -20º C for long-term storage. Under aseptic conditions, this product remains stable at 4°C until the CoA-listed retest date.

      Helpful?

  2. How should we store this reagent?

    1 answer
    1. As stated in the properties section of the product page, the storage temperature is 2-8°C

      Helpful?

Reviews

No rating value

Active Filters