Skip to Content
Merck
All Photos(2)

Key Documents

AAWG047S6

Millipore

S-Pak® Membrane Filter

mixed cellulose esters (MCE), pore size 0.8 μm, filter diam. 47 mm, white gridded membranes for microbiological analysis of beverages

Synonym(s):

Membrane filters, Semi-permeable membranes, S-Pak Filters 0.8µm 47mm white gridded

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
eCl@ss:
32031602
NACRES:
NB.24
pore size:
0.8 μm pore size
material:
gridded filter
mixed cellulose esters (MCE)
white filter
feature:
easily wetted with water
filter diam.:
47 mm
sterility:
sterile; ethylene oxide treated

material

gridded filter
mixed cellulose esters (MCE)
white filter

Agency

ISO 7704

sterility

sterile; ethylene oxide treated

feature

easily wetted with water

manufacturer/tradename

S-Pak®

filter diam.

47 mm

pore size

0.8 μm pore size

input

sample type beverage(s)
sample type wine
sample type bottled water
sample type sport drink
sample(s) (environmental)
sample type soft drink
sample type pharmaceutical(s)
sample type cider
sample type beer
sample type food(s)
sample type cosmetics

application(s)

bioburden testing
clinical
cosmetics
environmental
food and beverages
industrial qc
pharmaceutical

compatibility

for use with Microfil® Filtration Unit

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

S-Pak membrane Filters are made of Mixed cellulose Esters (MCE) with 0.8 μm pore size, white, gridded, 47mm diameter. These sterile, ready to use, individually packed filters are easy to use with no toxicity and hydrophobic spots. These filters are used to recover bacteria and other microorganisms from beverages, water and environment.

Application

Membrane Filtration method for Microbial Analysis of beverages such as water, wine, beer, sport drinks.

Packaging

Individually sealed, with blue separator paper, sterile

Legal Information

MICROFIL is a registered trademark of Merck KGaA, Darmstadt, Germany
S-PAK is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Irrit. 2 - Flam. Sol. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

S-Pak® membrane filters for microbial bioburden testing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service