Skip to Content
Merck
All Photos(1)

Key Documents

8.21258

Sigma-Aldrich

2-Methyl-1,3-butadiene

(stabilised) for synthesis

Synonym(s):

2-Methyl-1,3-butadiene, Isoprene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHC(CH3)=CH2
CAS Number:
Molecular Weight:
68.12
MDL number:
UNSPSC Code:
12352002
EC Index Number:
201-143-3
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

605 hPa ( 20 °C)

Quality Level

Assay

≥98.0% (GC)

form

liquid

autoignition temp.

220 °C

potency

>2000 mg/kg LD50, oral (Rat)

expl. lim.

1-9.7 % (v/v)

bp

34 °C/1013 hPa

mp

-146 °C

transition temp

flash point -54 °C

density

0.68 g/cm3 at 20 °C

SMILES string

CC(=C)C=C

InChI

1S/C5H8/c1-4-5(2)3/h4H,1-2H2,3H3

InChI key

RRHGJUQNOFWUDK-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.681 - 0.683
Identity (IR): passes test

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - Flam. Liq. 1 - Muta. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-65.2 °F - closed cup

Flash Point(C)

-54 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service