Skip to Content
Merck
All Photos(1)

Key Documents

133302

Sigma-Aldrich

Cyclohexyl isocyanide

98%

Synonym(s):

Isocyanocyclohexane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11NC
CAS Number:
Molecular Weight:
109.17
Beilstein:
3662332
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.45 (lit.)

density

0.878 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

[C-]#[N+]C1CCCCC1

InChI

1S/C7H11N/c1-8-7-5-3-2-4-6-7/h7H,2-6H2

InChI key

XYZMOVWWVXBHDP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cyclohexyl isocyanide reacts with dimethyl acetylenedicarboxylate to give a mixture of cyclopenta[b]pyridine derivatives, azaspirononatriene derivative and the azabicyclononatriene. It reacts with dialkyl acetylenedicarboxylates to form 1:1 intermediate which on facile addition to 1-benzylisatin and tryptantrin yields highly functionalized novel unsaturated γ-spiroiminolactones.

Application

  • Isocyanide in coordination chemistry: The study on mixed "2 + 1" tricarbonyl dithiocarbamate complexes highlights Cyclohexyl isocyanide′s role as an effective monodentate ligand, contributing to advancements in radiopharmaceutical applications using Re, Tc, and Re isotopes (Shegani et al., 2021).
  • Organic synthesis reagent in vascular treatments: The article discusses the use of Cyclohexyl isocyanide in modifying the endocannabinoid system, emphasizing its potential in developing treatments for conditions like hypertension through biochemical pathway modulation (Baranowska-Kuczko et al., 2021).
  • Chemical process optimization in dye decolorization: Cyclohexyl isocyanide plays a crucial role in the covalent immobilization of enzymes used for the decolorization of textile dyes, demonstrating its utility in environmental chemistry and industrial applications related to pollution control (Salami et al., 2018).
  • Application in nanocellulose modification: Demonstrates the versatility of Cyclohexyl isocyanide in nanotechnology by facilitating the covalent attachment of temperature-responsive polymers to cellulose nanofibrils, enhancing the material′s properties for use in smart textiles and responsive materials (Khine et al., 2018).

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

170.6 °F - closed cup

Flash Point(C)

77 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure and Thermal Isomerization of the Adducts Formed in the Reaction of Cyclohexyl Isocyanide with Dimethyl Acetylenedicarboxylate1.
Junjappa H, et al.
The Journal of Organic Chemistry, 63(26), 9801-9805 (1998)
Synthesis of some novel ?-spiroiminolactones from reaction of cyclohexyl isocyanide and dialkyl acetylene dicarboxylates with 1-benzylisatin and tryptanthrine.
Azizian J, et al.
Synthetic Communications, 33(3), 387-391 (2003)
Ghasem Marandi
Combinatorial chemistry & high throughput screening, 21(4), 298-301 (2018-04-18)
The reaction of cyclohexylisocyanide and 2-aminopyridine-3- carboxylic acid in the presence of benzaldehyde derivatives in ethanol led to 3-(cyclohexylamino)-2- arylimidazo[1,2-a]pyridine-8-carboxylic acids in high yields. In a three component condensation reaction, isocyanide reacts with 2-aminopyridine-3-carboxylic acid and aromatic aldehydes without any
Sodam Kim et al.
Journal of controlled release : official journal of the Controlled Release Society, 322, 337-345 (2020-04-05)
Nuclear medicine is a routine but essential clinical option for diagnostic imaging and disease treatment. Encapsulating radioisotopes in injectable biodegradable hydrogels is ideal for localizing radiation sources to target tissues or organs to achieve long-term, low-dose radiotherapy. However, difficulties in
Marius Brehm et al.
Small (Weinheim an der Bergstrasse, Germany), 16(10), e1905971-e1905971 (2020-01-28)
Miniaturization and parallelization of combinatorial organic synthesis is important to accelerate the process of drug discovery while reducing the consumption of reagents and solvents. This work presents a miniaturized platform for on-chip solid-phase combinatorial library synthesis with UV-triggered on-chip cell

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service