Skip to Content
Merck
All Photos(2)

Key Documents

8.21093

Sigma-Aldrich

2-Methyltetrahydrofuran

(stabilised) for synthesis

Synonym(s):

2-MeTHF, Tetrahydro-2-methylfuran, Tetrahydrosilvan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H10O
CAS Number:
Molecular Weight:
86.13
Beilstein:
102448
MDL number:
UNSPSC Code:
12352005
EC Index Number:
202-507-4
NACRES:
NA.22
bp:
78-80 °C (lit.)
vapor pressure:
136 hPa ( 20 °C)
Pricing and availability is not currently available.

vapor pressure

136 hPa ( 20 °C)

Quality Level

Assay

≥98% (GC)

form

liquid

autoignition temp.

270 °C

potency

300-2000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

expl. lim.

1.2-5.7 % (v/v)

refractive index

n20/D 1.406 (lit.)

bp

78-80 °C (lit.)

mp

-136 °C

transition temp

flash point -11 °C

solubility

150 g/L

density

0.86 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

CC1CCCO1

InChI

1S/C5H10O/c1-5-3-2-4-6-5/h5H,2-4H2,1H3

InChI key

JWUJQDFVADABEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Production of microalgal-based carbon encapsulated iron nanoparticles (ME-nFe) to remove heavy metals in wastewater: This study explores the application of ammonium iron(III) sulfate dodecahydrate in the synthesis of microalgal-based carbon encapsulated iron nanoparticles. These nanoparticles are used for the efficient removal of heavy metals from wastewater, presenting a significant advancement in environmental cleanup technologies (Mantovani et al., 2023).

  • pH responsive controlled release of anti-cancer hydrophobic drugs from sodium alginate and hydroxyapatite bi-coated iron oxide nanoparticles: This research utilizes ammonium iron(III) sulfate dodecahydrate in the development of sodium alginate and hydroxyapatite bi-coated iron oxide nanoparticles. These nanoparticles are designed for the pH-responsive controlled release of hydrophobic anti-cancer drugs, offering potential enhancements in targeted cancer therapy (Manatunga et al., 2017).

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.852 - 0.855
Peroxide (as H₂O₂): ≤ 0.005 %
Identity (IR): passes test
The material is stabilised with Butylhydroxytoluene (BHT).
At time of analytical testing a content of >100 ppm stabiliser was determined.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

14.0 °F - closed cup

Flash Point(C)

-10.0 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service