Skip to Content
Merck
All Photos(1)

Key Documents

8.20670

Sigma-Aldrich

Dimethylarsinic acid sodium salt trihydrate

for synthesis

Synonym(s):

Dimethylarsinic acid sodium salt trihydrate, Dimethylarsinic acid sodium salt, Cacodylic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H6AsNaO2 · 3H2O
CAS Number:
Molecular Weight:
214.03
MDL number:
UNSPSC Code:
12352107
EC Index Number:
204-708-2
NACRES:
NA.22
grade:
for synthesis
form:
solid
solubility:
2000 g/L
Pricing and availability is not currently available.

grade

for synthesis

Quality Level

form

solid

potency

>2000 mg/kg LD50, skin (Rabbit)

pH

9.0-9.5 (20 °C, 100 g/L in H2O)

mp

60 °C

solubility

2000 g/L

bulk density

870 kg/m3

storage temp.

2-30°C

SMILES string

[As](=O)([O-])(C)C.[Na+]

InChI

1S/C2H7AsO2.Na/c1-3(2,4)5;/h1-2H3,(H,4,5);/q;+1/p-1

InChI key

IHQKEDIOMGYHEB-UHFFFAOYSA-M

Analysis Note

Assay (HClO₄): ≥ 97.0 %
Identity (IR): passes test

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B

Storage Class Code

6.1A - Combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service