Skip to Content
Merck
All Photos(1)

Documents

20835

Sigma-Aldrich

Cacodylic acid

≥99.0%

Synonym(s):

Dimethylarsinic acid, Dimethylarsonic acid, Hydroxydimethylarsine oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2As(O)OH
CAS Number:
Molecular Weight:
138.00
Beilstein:
1736965
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0%

form

solid

reaction suitability

core: arsenic
reagent type: catalyst

SMILES string

C[As](C)(O)=O

InChI

1S/C2H7AsO2/c1-3(2,4)5/h1-2H3,(H,4,5)

InChI key

OGGXGZAMXPVRFZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Used as herbicides.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erik J Tokar et al.
Toxicology letters, 209(2), 179-185 (2012-01-11)
Inorganic arsenic, an early life carcinogen in humans and mice, can initiate lesions promotable by other agents in later life. The biomethylation product of arsenic, dimethylarsinic acid (DMA), is a multi-site tumor promoter. Thus, pregnant CD1 mice were given drinking
S Yamamoto et al.
Mutation research, 386(3), 353-361 (1997-06-01)
The modifying effects of dimethylarsinic acid (DMA), the major metabolite of ingested arsenicals in most mammals, on chemical carcinogenesis were investigated using rat in vivo models and reviewed here. In a multi-organ bioassay, rats pretreated with 5 carcinogens were administered
Tomohiro Narukawa et al.
Journal of agricultural and food chemistry, 60(4), 1122-1127 (2012-01-10)
A monitoring test for arsenic species in white rice flour was developed and applied to flours made from 20 samples of polished rice collected from locations all over Japan. The arsenic species in white rice flour made from five samples
Hua Naranmandura et al.
Toxicology and applied pharmacology, 260(3), 241-249 (2012-03-20)
The purpose of present study was to characterize the endoplasmic reticulum stress and generation of ROS in rat liver RLC-16 cells by exposing to trivalent dimethylarsinous acid (DMAIII) and compared with that of trivalent arsenite (iAsIII) and monomethylarsonous acid (MMAIII).
Hua Naranmandura et al.
Analytical and bioanalytical chemistry, 405(6), 1885-1891 (2012-06-27)
Arsenic (+3 oxidation state) methyltransferase (As3mt) plays a central role in the enzymatically catalyzed conversion of inorganic arsenic into methylated metabolites. Most studies of the metabolism and disposition of arsenicals following exposure to inorganic arsenic focus on the formation and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service