Skip to Content
Merck

Skip To

8.20171

Bromine

for synthesis

Synonym(s):

Bromine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
Molecular Weight:
159.81
UNSPSC Code:
12352300
EC Index Number:
231-778-1
NACRES:
NA.22
MDL number:
Form:
liquid
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

233 hPa ( 20 °C)

Quality Segment

form

liquid

reaction suitability

reagent type: oxidant

kinematic viscosity

0.314 cSt(20 °C)

bp

58.8 °C/1013 hPa

mp

-7.3 °C

solubility

35 g/L

density

3.12 g/cm3 at 20 °C (liquid)

storage temp.

15-25°C

SMILES string

[Br-]

InChI

1S/BrH/h1H/p-1

InChI key

CPELXLSAUQHCOX-UHFFFAOYSA-M

General description

for synthesis

Application

Bromine can be used as an oxidizing agent for the oxidation of:
  • Primary alcohols to either aldehydes or esters and secondary alcohols to give ketones.
  • Alcohols or diols to tetrahydrofurans in the presence of silver(I) salts.
  • Enediol bis-trimethylsilyl ethers to α-diketones.
  • Aldehydes to esters in the presence of alcohol solvents and sodium bicarbonate buffer.
  • Aliphatic and aromatic thiols to disulfides.

Bromine can also be used along with other co-reactants such as hexamethylphosphoric triamide (HMPA) and bis(tributyltin) oxide (HBD) to selectively oxidize secondary alcohols. However, bromine/nickel carboxylates convert 1,4-diols to γ-butyrolactones by selective oxidation of the primary alcohols.

Analysis Note

Assay (iodometric)≥ 99.0 %AppearanceBrownish-red fuming liquid

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.019451.019471.01948
description

for synthesis

description

extra pure

description

99.9999 Suprapur®

description

for analysis EMSURE® ACS,ISO,Reag. Ph Eur

Quality Level

200

Quality Level

200

Quality Level

100

Quality Level

300

solubility

35 g/L

solubility

35 g/L

solubility

35 g/L

solubility

35 g/L

reaction suitability

reagent type: oxidant

reaction suitability

-

reaction suitability

-

reaction suitability

-

bp

58.8 °C/1013 hPa

bp

58.8 °C/1013 hPa

bp

58.8 °C/1013 hPa

bp

58.8 °C/1013 hPa

density

3.12 g/cm3 at 20 °C (liquid)

density

3.12 g/cm3 at 20 °C (liquid)

density

3.12 g/cm3 at 20 °C (liquid)

density

3.12 g/cm3 at 20 °C (liquid)

storage temp.

15-25°C

storage temp.

15-25°C

storage temp.

15-25°C

storage temp.

15-25°C


Still not finding the right product?

Explore all of our products under Bromine


signalword

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1A

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

Storage Class

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters