Skip to Content
Merck
All Photos(1)

Key Documents

1.01948

Supelco

Bromine

for analysis EMSURE® ACS,ISO,Reag. Ph Eur

Synonym(s):

Bromine

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
159.81
MDL number:
UNSPSC Code:
41116105
EC Index Number:
231-778-1
NACRES:
NA.21
Pricing and availability is not currently available.

grade

ACS reagent

Quality Level

Agency

reag. ISO
reag. Ph. Eur.

vapor pressure

233 hPa ( 20 °C)

product line

EMSURE®

form

liquid

kinematic viscosity

0.314 cSt(20 °C)

bp

58.8 °C/1013 hPa

mp

-7.3 °C

solubility

35 g/L

density

3.12 g/cm3 at 20 °C (liquid)

storage temp.

15-25°C

SMILES string

[Br-]

InChI

1S/BrH/h1H/p-1

InChI key

CPELXLSAUQHCOX-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

for analysis EMSURE® ACS,ISO,Reag. Ph Eur

Application

Bromine for analysis EMSURE® ACS,ISO,Reag. Ph Eur. CAS 7726-95-6, molar mass 159.81 g/mol.

Analysis Note

Assay≥ 99.5 %Chlorine (Cl)≤ 500 ppmIodine (I)≤ 10 ppmHeavy metals (as Pb)≤ 2 ppmSulfur compounds (as SO4)≤ 5 ppmAl (Aluminium)≤ 0.050 ppmBa (Barium)≤ 0.100 ppmBe (Beryllium)≤ 0.020 ppmBi (Bismuth)≤ 0.100 ppmCa (Calcium)≤ 0.200 ppmCd (Cadmium)≤ 0.050 ppmCo (Cobalt)≤ 0.020 ppmCr (Chromium)≤ 0.050 ppmCu (Copper)≤ 0.100 ppmFe (Iron)≤ 0.500 ppmGe (Germanium)≤ 0.050 ppmK (Potassium)≤ 0.100 ppmLi (Lithium)≤ 0.020 ppmMg (Magnesium)≤ 0.500 ppmMn (Manganese)≤ 0.020 ppmMo (Molybdenum)≤ 0.020 ppmNa (Sodium)≤ 0.500 ppmNi (Nickel)≤ 0.020 ppmSr (Strontium)≤ 0.020 ppmTi (Titanium)≤ 0.100 ppmTl (Thallium)≤ 0.050 ppmV (Vanadium)≤ 0.050 ppmZn (Zinc)≤ 0.100 ppmZr (Zirconium)≤ 0.100 ppmOrganic bromine compoundspasses testNon volatile matter≤ 50 ppmDensity (d 20/20): about 3.1 g/cm³

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Aquatic Acute 1 - Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

6.1B - Non-combustible, acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service