Skip to Content
Merck
All Photos(2)

Key Documents

8.16004

Sigma-Aldrich

(S)-(+)-Glutamic acid

for synthesis

Synonym(s):

(S)-(+)-Glutamic acid, L-α-Aminoglutaric acid, Glu, L-alpha-Aminoglutaric acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO4
CAS Number:
Molecular Weight:
147.13
MDL number:
UNSPSC Code:
12352209
EC Index Number:
200-293-7
NACRES:
NA.22

grade

for synthesis
synthesis grade

Quality Level

Assay

≥99% (acidimetric)

form

solid

potency

>30000 mg/kg LD50, oral (Rat)

pH

3.0-3.5 (25 °C, 8.6 g/L in H2O)

mp

160 °C

solubility

11.1 g/L

density

1.54 g/cm3 at 20 °C

bulk density

460 kg/m3

storage temp.

2-30°C

SMILES string

[N+H3]C(CCC(=O)O)C(=O)[O-]

InChI

1S/C5H9NO4/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H,7,8)(H,9,10)

InChI key

WHUUTDBJXJRKMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Analysis Note

Assay (acidimetric): ≥ 99.0 %
Spec. rotation [α²0/D (c=10 in HCl 2 mol/l): +30.0 - +32.5 °
Identity (IR): passes test

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service