Skip to Content
Merck
All Photos(1)

Key Documents

8.09711

Sigma-Aldrich

Methyl acetate

for synthesis

Synonym(s):

Methyl acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6O2
CAS Number:
Molecular Weight:
74.08
MDL number:
UNSPSC Code:
12352108
EC Index Number:
201-185-2
NACRES:
NA.22
Assay:
≥99% (GC)
bp:
56-58 °C/1013 hPa
vapor pressure:
217 hPa ( 20 °C)
Pricing and availability is not currently available.

vapor pressure

217 hPa ( 20 °C)

Quality Level

Assay

≥99% (GC)

form

liquid

autoignition temp.

455 °C

potency

>5000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rat)

expl. lim.

3.1-16 % (v/v)

pH

7

bp

56-58 °C/1013 hPa

mp

-98 °C

transition temp

flash point -13 °C

solubility

250 g/L

density

0.93 g/cm3 at 20 °C

storage temp.

no temp limit

SMILES string

O(C)C(=O)C

InChI

1S/C3H6O2/c1-3(4)5-2/h1-2H3

InChI key

KXKVLQRXCPHEJC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Adaptable heat pump-assisted dividing-wall column design for intensified downstream processing of bio-propionic acid: This article describes the esterification and separation processes in which methyl acetate is used to achieve higher purity levels in chemical production. This method enhances the recovery of methyl acetate at a purity of 99.9% (T Janković, AJJ Straathof, AA Kiss - Separation and Purification Technology, 2024 - Elsevier) (Source).
  • Beta-irradiation of biomass in the presence of 1-alkyl-3-methyl-imidazolium ionic liquids: Discusses the unexpected product formation during the beta-irradiation of biomass, where methyl acetate plays a crucial role in the chemical reactions of biomass conversion processes (AF Lehrhofer, M Bacher, H Hettegger, S Böhmdorfer - Cellulose, 2024 - Springer) (Source).
  • Supramolecular Gels Based on C3-Symmetric Amides: Explores the application of methyl acetate in the formation of supramolecular gels aimed at environmental applications such as anion-sensing and dye removal from water, demonstrating its versatility in advanced material science (G Kuppadakkath, SS Jayabhavan, KK Damodaran - Molecules, 2024 - mdpi.com) (Source).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Density (d 20 °C/ 4 °C): 0.932 - 0.934
Identity (IR): passes test

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

8.6 °F - closed cup

Flash Point(C)

-13 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service