Skip to Content
Merck
All Photos(1)

Key Documents

8.07572

Sigma-Aldrich

Pyrazole

for synthesis

Synonym(s):

Pyrazole, 1,2-Diazol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H4N2
CAS Number:
Molecular Weight:
68.08
MDL number:
UNSPSC Code:
12352005
EC Index Number:
206-017-1
NACRES:
NA.22

Quality Level

Assay

≥99.0% (GC)

form

crystals

potency

1010 mg/kg LD50, oral (Rat)

mp

66-68 °C

storage temp.

2-30°C

SMILES string

[nH]1nccc1

InChI

1S/C3H4N2/c1-2-4-5-3-1/h1-3H,(H,4,5)

InChI key

WTKZEGDFNFYCGP-UHFFFAOYSA-N

Related Categories

Application


  • Uranium extraction with pyrazole-based polymers: A study demonstrated the capability of a pyrazole-based porous organic polymer to extract uranium, offering potential applications in environmental cleanup and nuclear waste management (Yu et al., 2024).

  • Pyrazole derivatives for agricultural fungicides: Novel pyrazole-4-carboxamide derivatives containing an oxime ether group were synthesized and evaluated as potential succinate dehydrogenase inhibitors (SDHIs) for controlling Rhizoctonia solani in crops, suggesting their utility in agriculture (Luo et al., 2024).

  • Pyrazole-based fluorescent probes: Research on the synthesis and application of specific N(2)H(4) fluorescent probes based on the pyrazole structure demonstrated their utility with aggregation-induced emission (AIE) properties, highlighting potential applications in bioimaging (Yang et al., 2024).

Analysis Note

Assay (GC, area%): ≥ 99.0 % (a/a)
Melting range (lower value): ≥ 66 °C
Melting range (upper value): ≤ 69 °C
Identity (IR): passes test

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2 - STOT RE 2

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service