Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C5H8N2
CAS Number:
Molecular Weight:
96.13
NACRES:
NA.22
PubChem Substance ID:
eCl@ss:
39160503
UNSPSC Code:
12352100
EC Number:
200-657-5
MDL number:
Beilstein/REAXYS Number:
106325
Assay:
≥99.0% (GC)
Quality Segment
assay
≥99.0% (GC)
manufacturer/tradename
Wacker Chemie AG
bp
218 °C (lit.)
mp
105-108 °C (lit.)
SMILES string
Cc1cc(C)[nH]n1
InChI
1S/C5H8N2/c1-4-3-5(2)7-6-4/h3H,1-2H3,(H,6,7)
InChI key
SDXAWLJRERMRKF-UHFFFAOYSA-N
Still not finding the right product?
Explore all of our products under 3,5-Dimethylpyrazole
signalword
Warning
hcodes
Hazard Classifications
Acute Tox. 4 Oral - STOT RE 2
target_organs
Liver
Storage Class
13 - Non Combustible Solids
wgk
WGK 3
ppe
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

