8.01453
Bis(2-ethylhexyl) adipate
for synthesis
Synonym(s):
Bis(2-ethylhexyl) adipate, Bis(2-ethylhexyl) hexanedioate, Di-(2-ethylhexyl) adipate, Diisooctyl adipate, Hexanedioic bis(2-ethylhexyl ester)
About This Item
Recommended Products
vapor pressure
<0.001 hPa ( 20 °C)
Quality Level
Assay
≥98.0% (GC)
form
liquid
autoignition temp.
350 °C (DIN 51794)
potency
>5000 mg/kg LD50, oral (Rat)
8410 mg/kg LD50, skin (Rabbit)
expl. lim.
0.24 % (v/v)
pH
7 (20 °C in H2O)
bp
247-252 °C/27 hPa
mp
-68--66 °C
transition temp
flash point 195 °C
density
0.93 g/cm3 at 20 °C
storage temp.
2-30°C
SMILES string
O(CC(CCCC)CC)C(=O)CCCCC(=O)OCC(CCCC)CC
InChI
1S/C22H42O4/c1-5-9-13-19(7-3)17-25-21(23)15-11-12-16-22(24)26-18-20(8-4)14-10-6-2/h19-20H,5-18H2,1-4H3
InChI key
SAOKZLXYCUGLFA-UHFFFAOYSA-N
Related Categories
Application
- Influence on Poly(lactic acid) Fiber Production: Research highlights how low-molecular-weight esters like Bis(2-ethylhexyl) adipate can enhance the melt spinning and structural properties of poly(lactic acid) fibers, suggesting potential advancements in textile manufacturing technologies (Gzyra-Jagieła et al., 2024).
- Environmental Analysis of Atmospheric Particles: A new method using GC Orbitrap-MS has been developed for the simultaneous analysis of phthalate esters and Bis(2-ethylhexyl) adipate, it is commonly used as plasticisers in consumer and industrial products (Argamino et al., 2024).
- Study on Reproductive Endocrine Disruption: Examination of the effects of Bis(2-ethylhexyl) adipate, among other plasticizers, on gene expression related to the reproductive endocrine system in Japanese medaka fish, contributing to our understanding of environmental endocrine disruptors (Horie and Uaciquete, 2023).
- Health Risk Assessments in Urban Environments: Bis(2-ethylhexyl) adipate was included in a comprehensive study assessing health risks associated with organic compounds in atmospheric fine particles, aimed at improving public health safety measures in populated regions (Gao et al., 2024).
- Exposure from Plastic Water Bottles: The presence and implications of micro(nano)plastics, including polymers plasticized with Bis(2-ethylhexyl) adipate, in single-use plastic bottles were analyzed, indicating potential human exposure risks from everyday plastic use (Vega-Herrera et al., 2023).
Analysis Note
Density (d 20 °C/ 4 °C): 0.924 - 0.926
Identity (IR): passes test
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
384.8 °F - closed cup
Flash Point(C)
196 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service