Skip to Content
Merck
All Photos(1)

Key Documents

538140

Sigma-Aldrich

Trioctyl trimellitate

99%

Synonym(s):

TOTM, Tris(2-ethylhexyl) trimellitate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3-1,2,4-[CO2CH2CH(C2H5)(CH2)3CH3]3
CAS Number:
Molecular Weight:
546.78
Beilstein:
2683525
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99%

refractive index

n20/D 1.485 (lit.)

bp

414 °C (lit.)

density

0.99 g/mL at 25 °C (lit.)

SMILES string

CCCCC(CC)COC(=O)c1ccc(C(=O)OCC(CC)CCCC)c(c1)C(=O)OCC(CC)CCCC

InChI

1S/C33H54O6/c1-7-13-16-25(10-4)22-37-31(34)28-19-20-29(32(35)38-23-26(11-5)17-14-8-2)30(21-28)33(36)39-24-27(12-6)18-15-9-3/h19-21,25-27H,7-18,22-24H2,1-6H3

InChI key

KRADHMIOFJQKEZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Trioctyl trimellitate (TOTM) is a flammable plasticizer used to improve the flexibility and durability of poly(vinyl chloride) (PVC).

Application

TOTM can plasticize PVC to form a nanocomposite with multi-walled carbon nanotube which can be potentially used for anti-static applications.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

505.4 °F - closed cup

Flash Point(C)

263 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Morphology correlated free volume studies of multi-walled carbon nanotube plasticized poly (vinyl chloride) nanocomposites: Positronium probes and electrical properties
Francis E, et al.
Polymer, 141(27), 232-243 (2018)
Hierarchical nanoporous silica doped with tin as novel multifunctional hybrid material to flexible poly (vinyl chloride) with greatly improved flame retardancy and mechanical properties
Pan Y, et al.
Chemical Engineering Journal, 295(27), 451-460 (2016)
Structure and diffusion behavior of trioctyl trimellitate (TOTM) in PVC film studied by ATR-IR spectroscopy
Lai H, et al.
Industrial & Engineering Chemistry Research, 51(27), 9365-9375 (2012)
J R Hodgson
Toxicology and industrial health, 3(2), 49-61 (1987-06-01)
The results of these studies demonstrate that Tri(2-ethylhexyl)trimellitate (TOTM) produces the sam spectrum of morphological and biochemical changes in the rat liver as di(2-ethylhexyl)phthalate (DEHP). TOTM, however, was much less potent in its action, with a dietary level of 2.0%
V S Turner et al.
Vox sanguinis, 69(3), 195-200 (1995-01-01)
Platelet concentrates (PCs), stored for 5 days in PL 2209, a new polyvinyl chloride (PVC) storage container plasticised with butyryl trihexyl citrate, were compared with those stored in PL 1240, a PVC platelet container plasticised with triethylhexyl trimellitate. In part

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service