4.80112
Trifluoroacetic acid solution
0.1% (v:v) trifluoroacetic acid, suitable for LC/MS, for use with LICHROSOLV®
Synonym(s):
Water with 0.1% (v:v) trifluoroacetic acid
About This Item
LC/MS: suitable
Recommended Products
Quality Level
product line
LiChrosolv®
form
liquid
concentration
0.095-0.105 % (v/v)
technique(s)
HPLC: suitable
LC/MS: suitable
impurities
≤0.03 ppm K (Potassium)
≤0.03 ppm Mg (Magnesium)
≤0.2 ppm Ca (Calcium)
≤0.5 ppm Fe (Iron)
≤0.7 ppm Na (Sodium)
≤50 ppm Al (Aluminium)
color
APHA: ≤10
compatibility
for use with LICHROSOLV®
storage temp.
2-30°C
General description
Preparation Note
Analysis Note
Identity: passes test
Color: ≤ 10 Hazen
Al (Aluminium): ≤ 0.01 ppm
Ca (Calcium): ≤ 0.2 ppm
Fe (Iron): ≤ 0.5 ppm
K (Potassium): ≤ 0.03 ppm
Mg (Magnesium): ≤ 0.03 ppm
Na (Sodium): ≤ 0.7 ppm
Gradient grade (at 210 nm): ≤ 10.0 mAU
Gradient grade (at 254 nm): ≤ 2.0 mAU
Suitable for LC-MS; Intensity of background mass peak based on reserpine(APCI/ESI positive): ≤ 2 ppb
Filtered by 0.2µm filter
Other Notes
Legal Information
related product
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Learn the tips and tricks of selecting and preparing a suitable mobile phase buffer for your HPLC separations and method developments for samples that contain ionizable compounds.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service