443300
MCI-186
A free radical scavenger and antioxidant that reduces post-ischemic brain injury.
Synonym(s):
MCI-186, 3-Methyl-1-phenyl-2-pyrazolin-5-one, PMP, Edaravone
About This Item
Recommended Products
Quality Level
description
Merck USA index - 14, 6713
Assay
≥98% (HPLC)
form
solid
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
protect from light
color
cream
solubility
DMSO: 100 mg/mL
shipped in
ambient
storage temp.
2-8°C
InChI
1S/C10H10N2O/c1-8-7-10(13)12(11-8)9-5-3-2-4-6-9/h2-6H,7H2,1H3
InChI key
QELUYTUMUWHWMC-UHFFFAOYSA-N
General description
Biochem/physiol Actions
A free radical scavenger and antioxidant
Packaging
Warning
Reconstitution
Other Notes
Kawai, H., et al. 1997. J. Pharmacol. Exp. Ther. 281, 921.
Yamamoto, Y., et al. 1996. Redox. Report 2, 333.
Watanabe, T., et al. 1994. J. Pharmacol. Exp. Ther. 268, 1597.
Nishi, H., et al. 1989. Stroke 20, 1236.
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service