Skip to Content
Merck
All Photos(1)

Key Documents

14-338

Sigma-Aldrich

ROKα/ROCK-II Protein, active, rat, 10 µg

Active, rat, N-terminal, His-tagged, fusion protein corresponding to amino acids 2-543 of rat ROKa/ROCK-II. For use in Kinase Assays.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32

biological source

rat

Quality Level

mol wt

Mw 66 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

shipped in

dry ice

Gene Information

General description

N-terminal, His-tagged, fusion protein corresponding to amino acids 2-543 of rat ROKa/ROCK-II.

Note: The preparation contains numerous Sf21 cellular proteins which bind weakly to Ni-NTA agarose.
Product Source: Rat ROKa/ROCK-II expressed in Sf21 insect cells.

Biochem/physiol Actions

Protein Target: ROKα/ROCK-II
Target Sub-Family: AGC

Quality

Rroutinely evaluated using the S6 Kinase Assay Kit

Physical form

10μg of enzyme in 2.3μl of 50mM Tris/HCl pH7.5, 150mM NaCl, 0.1mM EGTA, 0.03% Brij-35, 270mM sucrose, 2mM benzamidine, 0.4mM PMSF, 0.1% 2-mercaptoethanol. Frozen solution.
Ni-NTA agarose

Storage and Stability

6 months at -70°C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander Khromov et al.
The Journal of biological chemistry, 284(32), 21569-21579 (2009-06-18)
The reversible regulation of myosin light chain phosphatase (MLCP) in response to agonist stimulation and cAMP/cGMP signals plays an important role in the regulation of smooth muscle (SM) tone. Here, we investigated the mechanism underlying the inhibition of MLCP induced
David A Carlson et al.
Cell chemical biology, 25(10), 1195-1207 (2018-07-24)
Sustained vascular smooth muscle hypercontractility promotes hypertension and cardiovascular disease. The etiology of hypercontractility is not completely understood. New therapeutic targets remain vitally important for drug discovery. Here we report that Pim kinases, in combination with DAPK3, regulate contractility and
M Eto et al.
The Journal of biological chemistry, 276(31), 29072-29078 (2001-06-09)
Histamine stimulus triggers inhibition of myosin phosphatase-enhanced phosphorylation of myosin and contraction of vascular smooth muscle. In response to histamine stimulation of intact femoral artery, a smooth muscle-specific protein called CPI-17 (for protein kinase C-potentiated inhibitory protein for heterotrimeric myosin
Vincent Sauzeau et al.
Methods in molecular biology (Clifton, N.J.), 1527, 213-218 (2017-01-25)
Increased arterial tone and the resulting rise in peripheral vascular resistance are major determinants of the elevated arterial pressure in hypertension. The RhoA/Rho kinase signaling pathways are now recognized as a major regulator of vascular smooth muscle contraction and arterial
Cassandra L Hubert et al.
Journal of biomolecular screening, 8(4), 399-409 (2003-10-22)
The Rho-associated coiled-coil-containing protein serine/threonine kinases ROCK-I and ROCK-II are thought to play a major role in cytoskeletal dynamics by serving as downstream effectors of the Rho/Rac family of cytokine- and growth factor-activated small GTPases. As such, the ROCK family

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service