1.13990
Spectromelt® C 10
Grinding and tabletting aid for X-ray fluorescence spectroscopy, for X-ray fluorescence analysis
About This Item
Recommended Products
Quality Level
form
solid
technique(s)
XRF: suitable
solubility
water: partially soluble
cation traces
Al: ≤0,001%
Ba: ≤ 0,001%
Ca: ≤ 0,001%
Co: ≤ 0,001%
Cr: ≤ 0,001%
Cu: ≤ 0,001%
Fe: ≤ 0,001%
K: ≤ 0,001%
Mn: ≤ 0,001%
Mo: ≤ 0,001%
Ni: ≤ 0,001%
Pb: ≤ 0,001%
Si: ≤ 0,001%
Sr: ≤ 0,001%
Ti: ≤ 0,001%
V: ≤ 0,001%
storage temp.
no temp limit
General description
Analysis Note
Al (Aluminium): ≤ 0,001
Ba (Barium): ≤ 0,001
Ca (Calcium): ≤ 0,001
Co (Cobalt): ≤ 0,001
Cr (Chromium): ≤ 0,001
Cu (Copper): ≤ 0,001
Fe (Iron): ≤ 0,001
K (Potassium): ≤ 0,001
Mn (Manganese): ≤ 0,001
Mo (Molybdenum): ≤ 0,001
Ni (Nickel): ≤ 0,001
Pb (Lead): ≤ 0,001
Si (Silicon): ≤ 0,001
Sr (Strontium): ≤ 0,001
Ti (Titanium): ≤ 0,001
V (Vanadium): ≤ 0,001
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Atomic spectroscopy techniques like AAS, AES, ICP-OES, and ICP-MS identify and quantify elemental composition in samples.
Atomic spectroscopy techniques like AAS, AES, ICP-OES, and ICP-MS identify and quantify elemental composition in samples.
Atomic spectroscopy techniques like AAS, AES, ICP-OES, and ICP-MS identify and quantify elemental composition in samples.
Atomic spectroscopy techniques like AAS, AES, ICP-OES, and ICP-MS identify and quantify elemental composition in samples.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service