Skip to Content
Merck
All Photos(2)

Key Documents

1.09973

Supelco

Hydrochloric acid solution

c(HCl) = 0.1 mol/L (1N), concentrate for preparing 1000 mL volumetric solution for titration, Titrisol®

Synonym(s):

Hydrochloric acid solution, Hydrogen chloride solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
HCl
CAS Number:
Molecular Weight:
36.46
MDL number:
UNSPSC Code:
12352301
NACRES:
NA.21
Pricing and availability is not currently available.

Product Name

Hydrochloric acid solution, for 1000 ml, c(HCl) = 0.1 mol/l (0.1 N) Titrisol®

Quality Level

product line

Titrisol®

form

liquid

reaction suitability

reaction type: Acid-base reactions

concentration

0.1 M

technique(s)

titration: suitable

pH

<1 (20 °C in H2O)

density

1.03 g/cm3 at 20 °C

storage temp.

15-25°C

SMILES string

Cl

InChI

1S/ClH/h1H

InChI key

VEXZGXHMUGYJMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Preparation of epoxy-functionalized hierarchically porous hybrid monoliths via free radical polymerization and application in HILIC enrichment of glycopeptides.: This study outlines the preparation of epoxy-functionalized monoliths using hydrochloric acid solutions for glycopeptide enrichment, contributing to advancements in proteomics and analytical chemistry (Ma et al., 2019).

  • Simultaneous analysis of nine aromatic amines in mainstream cigarette smoke using online solid-phase extraction combined with liquid chromatography-tandem mass spectrometry.: The research presents a method for analyzing aromatic amines in cigarette smoke, utilizing hydrochloric acid solutions for sample preparation, enhancing detection sensitivity in environmental studies (Zhang et al., 2017).

  • A simplified HPLC method for determination of tryptophan in some cereals and legumes.: This paper details a simplified HPLC method using hydrochloric acid solutions for tryptophan determination in cereals and legumes, improving food quality analysis techniques (Çevikkalp et al., 2016).

  • The microwave induced plasma with optical emission spectrometry (MIP-OES) in 23 elements determination in geological samples.: This study uses hydrochloric acid solutions in MIP-OES to determine elemental composition in geological samples, contributing to advancements in geochemical analysis (Niedzielski et al., 2015).

  • Revisiting methods for the determination of bioavailable metals in coastal sediments.: The research revisits analytical methods for determining bioavailable metals in sediments using hydrochloric acid solutions, enhancing environmental monitoring and pollution assessment (Peña-Icart et al., 2014).

Legal Information

Titrisol is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. What is the purity of this product?

    1 answer
    1. This product is not specifically tested for purity and trace analysis testing is not performed. However, the purity may be calculated based on the lot-specific concentration, product density, and HCl molecular weight, as demonstrated in the example below:
      Molarity (moles/L) = Density (g/L) ÷ Molecular Weight (g/mol) * Purity
      0.1mol/L = 1030g/L ÷ 36.46mol/L * Purity
      0.1mol/L = 28.2501mol/L * Purity
      Purity = (0.1mol/L ÷ 28.2501mol/L) x 100
      = 0.35%

      Helpful?

  2. Hi, in what size volumetric flask will the HCL 0.1mol/l(0.1N) Titrisol need to be made up in, please

    1 answer
    1. Each ampule is sufficient to prepare 1000 mL of 0.1N hydrochloric acid solution. This will require a 1 L or 1000 mL volumetric flask.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service