Skip to Content
Merck
All Photos(1)

Key Documents

1.07393

Sigma-Aldrich

Proteinase K

(from Tritirachium album) solution in Tris/HCl pH 7.5; 0.01 mol/l; 600 mAnson-U/ml; for molecular biology EC 3.4.21.14

Synonym(s):

ProK, native proteinase K solution, non-specific protease

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.51

Pricing and availability is not currently available.

Quality Level

form

liquid

pH

7.5 (25 °C in H2O, undiluted)

density

1.10 g/cm3 at 20 °C

storage temp.

2-8°C

General description

Proteinase K is an endopeptidase that belongs to the subtilisin family of proteinases. The polypeptide chain contains 278 amino acids with the catalytic triad Asp39, His69, Ser224.

Application

Proteinase K has been used:
  • to de-crosslink immunoprecipitated samples
  • to treat the poly-L-lysine coated slides of colon tissues for terminal deoxynucleotidyl transferase dUTP nick end labeling (TUNEL) assay
  • in in situ hybridization

Biochem/physiol Actions

Proteinase K catalyzes the hydrolysis of esters and peptide bonds. It is used with non-aqueous hydrated solvents for synthesizing peptides. Proteinase K is used to break the cross-linking that develops secondary to formalin fixation and expose the target sequence for primers and polymerase.

Analysis Note

Appearance (colour): colourless
Appearance (clearness): clear
Activity (hemoglobin; pH 7.5; 37 °C): ≥ 600 mAnsonU/ml
Spec. activity (calc. on protein): ≥ 40.0 mAnsonU/mg
DNases (Nicking activity; pBR 322; 6 h; 37 °C): not detectable
RNases (RNA; 2 h; 37°C): not detectable
Colony count (aerobic bacteria): ≤ 10 CFU/ml

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Chavagnat et al.
Applied and environmental microbiology, 65(7), 3001-3007 (1999-07-02)
The general aminopeptidase PepN from Streptococcus thermophilus A was purified to protein homogeneity by hydroxyapatite, anion-exchange, and gel filtration chromatographies. The PepN enzyme was estimated to be a monomer of 95 kDa, with maximal activity on N-Lys-7-amino-4-methylcoumarin at pH 7
María Isabel Navarro-Mendoza et al.
Current biology : CB, 29(22), 3791-3802 (2019-11-05)
Centromeres are rapidly evolving across eukaryotes, despite performing a conserved function to ensure high-fidelity chromosome segregation. CENP-A chromatin is a hallmark of a functional centromere in most organisms. Due to its critical role in kinetochore architecture, the loss of CENP-A

Articles

Proteinase K aids in molecular biology applications by digesting structural proteins, removing nucleases, and isolating intact genomic DNA.

Proteinase K aids in molecular biology applications by digesting structural proteins, removing nucleases, and isolating intact genomic DNA.

Proteinase K aids in molecular biology applications by digesting structural proteins, removing nucleases, and isolating intact genomic DNA.

Proteinase K aids in molecular biology applications by digesting structural proteins, removing nucleases, and isolating intact genomic DNA.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service