Skip to Content
Merck
All Photos(1)

Key Documents

1.05865

Supelco

Magnesium oxide

for analysis EMSURE®

Synonym(s):

Magnesium oxide, Magnesia usta

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
MgO
CAS Number:
Molecular Weight:
40.30
MDL number:
UNSPSC Code:
12352303
EC Index Number:
215-171-9
NACRES:
NA.21
Assay:
≥97% (complexometric)
form:
solid
Pricing and availability is not currently available.

Quality Level

product line

EMSURE®

Assay

≥97% (complexometric)

form

solid

impurities

≤0.1% Substances insoluble in hydrochloric acid
≤0.5% Substances soluble in water

loss

≤3% loss on ignition, 1000 °C

pH

10.3 (20 °C in H2O, saturated solution)

bp

3600 °C/1000 hPa

mp

2800 °C

density

3.58 g/cm3 at 25 °C

bulk density

100 kg/m3

anion traces

carbonate (CO32-): ≤1.5% (as CO2)
chloride (Cl-): ≤0.01%
sulfate (SO42-): ≤0.02%

cation traces

Ba + Sr (as Ba): ≤0.005%
Ca: ≤0.10%
Cu: ≤0.001%
Fe: ≤0.005%
K: ≤0.005%
Na: ≤0.30%
Pb: ≤0.001%
Zn: ≤0.0005%

storage temp.

no temp limit

SMILES string

[Mg]=O

InChI

1S/Mg.O

InChI key

CPLXHLVBOLITMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Aluminosilicate-Supported Catalysts for the Synthesis of Cyclic Carbonates by Reaction of CO(2) with the Corresponding Epoxides.: Although not directly involving magnesium oxide, this research within analytical chemistry develops related catalytic processes which could cross-apply (Atzori et al., 2022).

Analysis Note

Assay (complexometric): ≥ 97 %
Substances soluble in water: ≤ 0.5 %
Substances insoluble in hydrochloric acid: ≤ 0.10 %
Carbonate (as CO₂): ≤ 1.5 %
Chloride (Cl): ≤ 0.01 %
Sulfate (SO₄): ≤ 0.02 %
Total nitrogen (N): ≤ 0.002 %
Ca (Calcium): ≤ 0.10 %
Cu (Copper): ≤ 0.001 %
Fe (Iron): ≤ 0.005 %
K (Potassium): ≤ 0.005 %
Na (Sodium): ≤ 0.30 %
Pb (Lead): ≤ 0.001 %
Zn (Zinc): ≤ 0.0005 %
Barium and Strontium (as Ba): ≤ 0.005 %
Loss on ignition (1000 °C): ≤ 3.0 %

Legal Information

EMSURE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service