Skip to Content
Merck
All Photos(1)

Key Documents

1.05862

SAFC

Magnesium oxide

light extra pure, Ph. Eur., BP, USP, E 530

Pharma Manufacturing

Synonym(s):

Magnesium oxide, Magnesia usta

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
MgO
CAS Number:
Molecular Weight:
40.30
MDL number:
UNSPSC Code:
12352303
E Number:
E530
EC Index Number:
215-171-9
Pricing and availability is not currently available.

Agency

BP
Ph. Eur.
USP

Quality Level

form

solid

pH

10.3 (20 °C in H2O, saturated solution)

bp

3600 °C/1000 hPa

mp

2800 °C

density

3.58 g/cm3 at 25 °C

application(s)

pharma/biopharma processes

storage temp.

15-25°C

SMILES string

[Mg]=O

InChI

1S/Mg.O

InChI key

CPLXHLVBOLITMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We offer a range of high quality critical raw materials plus the documentation and expertise to help our customers meet stringent regulatory requirements. Our SAFC® portfolio of high-quality products withstands strict quality control procedures.

Application

Magnesium oxide can be used as pH adjuster and is suitable for use in pharmaceutical processes.

Legal Information

SAFC is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service