Skip to Content
Merck
All Photos(3)

Key Documents

1.04360

Supelco

n-Heptane

greener alternative

for gas chromatography ECD and FID SupraSolv®

Synonym(s):

Heptane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH3
CAS Number:
Molecular Weight:
100.20
Beilstein:
1730763
MDL number:
UNSPSC Code:
41116105
EC Index Number:
205-563-8
NACRES:
NA.03
technique(s):
gas chromatography (GC): suitable
bp:
98 °C (lit.)
vapor pressure:
40 mmHg ( 20 °C)
83 mmHg ( 37.7 °C)
Pricing and availability is not currently available.

vapor density

3.5 (vs air)

Quality Level

vapor pressure

40 mmHg ( 20 °C)
83 mmHg ( 37.7 °C)

product line

SupraSolv®

Assay

≥99.5% (GC)

form

liquid

autoignition temp.

433 °F

potency

>5000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

expl. lim.

7 %

greener alternative product characteristics

Safer Solvents and Auxiliaries
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

gas chromatography (GC): suitable

refractive index

n20/D 1.387 (lit.)

bp

98 °C (lit.)

mp

−91 °C (lit.)

transition temp

flash point -4 °C

density

0.684 g/mL at 25 °C (lit.)

greener alternative category

storage temp.

2-30°C

SMILES string

CCCCCCC

InChI

1S/C7H16/c1-3-5-7-6-4-2/h3-7H2,1-2H3

InChI key

IMNFDUFMRHMDMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

SupraSolv® ECD and FID is specially developed for gas chromatography in combination with ECD (Electron Capture Detector) and FID (Flame Ionization detector). SupraSolv® solvents offer the largest specified retention time range, a clear baseline and a minimum signal-to-noise ratio leading to reliable and reproducible analysis results.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. Heptane is an environmentally preferable solvent compared to hexane and thus align with "Safer Solvents and Auxiliaries". Click here for more information.

Application

Appropriate for GC applications including sensitive detection in residue and environmental analysis.

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

SUPRASOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Questions

  1. What is the pack size of this n-Heptane

    1 answer
    1. This product is currently supplied in a 1 Liter and 2.5 Liter pack size.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service