Skip to Content
Merck
All Photos(3)

Key Documents

1.04390

Supelco

n-Heptane

greener alternative

for liquid chromatography LiChrosolv®

Synonym(s):

Heptane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH3
CAS Number:
Molecular Weight:
100.20
Beilstein:
1730763
MDL number:
UNSPSC Code:
12191502
EC Index Number:
205-563-8
NACRES:
NA.03
Assay:
≥99.3% (GC)
technique(s):
HPLC: suitable
bp:
98 °C (lit.)
vapor pressure:
40 mmHg ( 20 °C)
83 mmHg ( 37.7 °C)
Pricing and availability is not currently available.

vapor density

3.5 (vs air)

Quality Level

vapor pressure

40 mmHg ( 20 °C)
83 mmHg ( 37.7 °C)

product line

LiChrosolv®

grade

isocratic

Assay

≥99.3% (GC)

form

liquid

autoignition temp.

433 °F

potency

>5000 mg/kg LD50, oral (Rat)
>2000 mg/kg LD50, skin (Rabbit)

expl. lim.

7 %

greener alternative product characteristics

Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

technique(s)

HPLC: suitable

impurities

≤0.0002 meq/g Acidity
≤0.0002 meq/g Alkalinity
≤0.005% Water

evapn. residue

≤2.0 mg/L

transmittance

210 nm, ≥50%
220 nm, ≥80%
245 nm, ≥98%

refractive index

n20/D 1.387 (lit.)

bp

98 °C (lit.)

mp

−91 °C (lit.)

transition temp

flash point -4 °C

solubility

0.05 g/L

density

0.684 g/mL at 25 °C (lit.)

greener alternative category

storage temp.

2-30°C

SMILES string

CCCCCCC

InChI

1S/C7H16/c1-3-5-7-6-4-2/h3-7H2,1-2H3

InChI key

IMNFDUFMRHMDMM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

LiChrosolv® solvents exhibit high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level. This makes them suitable for reproducible separations.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. Heptane is an environmentally preferable solvent compared to hexane and thus align with "Safer Solvents and Auxiliaries". Click here for more information.

Application

Intended for U/HPLC and LC-MS/MS applications.[1]
Also reported for use as a component of extraction mixture in screening drugs of abuse in urine samples by GC-MS.[2]

Features and Benefits

  • suitability tested and specified for UHPLC-MS and UHPLC-UV: for analytical flexibility
  • specified quality in positive and negative electrospray ionization (ESI) and atmospheric-pressure chemical ionization-mass spectrometry (APCI-MS) for lowest detection limits and confidence in analyses in all important MS modes (test 1)
—ESI/APCI (+) < 2 ppb
—ESI/APCI (-) < 10 ppb
  • lowest impurity profile: for interference-free baselines (test 2)
  • microfiltered through 0.2 μm filter (test 3) to provide:
—a long lifespan to filters and mechanical parts of HPLC systems
—reduced risk of column clogging
  • packaged in borosilicate glass bottles for minimized contamination with metal ions
  • lowest levels of trace metal impurities to minimize metal ion adduct formation
—<5ppb
  • lowest level of polyethylene glycol (PEG) impurities in UHPLC-MS solvent lineup (PEG S/N signal-to-noise-ratio < 50)

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Purity (GC): ≥ 99.3 %
Identity (IR): conforms
Evaporation residue: ≤ 2.0 mg/l
Water: ≤ 0.005 %
Acidity: ≤ 0.0002 meq/g
Alkalinity: ≤ 0.0002 meq/g
Transmission (at 210 nm): ≥ 50 %
Transmission (at 220 nm): ≥ 80 %
Transmission (from 245 nm): ≥ 98 %
Filtered by 0.2 µm filter

Other Notes

Explore various lab safety accessories and equipment for safe handling of solvents to increase your safety level from the first usage.

Legal Information

LICHROSOLV is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

24.8 °F - closed cup

Flash Point(C)

-4 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

LC-MS/MS with atmospheric pressure chemical ionisation to study the effect of UV treatment on the formation of vitamin D3 and sterols in plants
Japelt RB, et al.
Food Chemistry, 129(1), 217-225 (2011)
Intoxication caused by new psychostimulants: analytical methods to disclose acute and chronic use of benzofurans and ethylphenidate
Barcelo B, et al.
International Journal of Legal Medicine, 131(6), 1543-1553 (2017)

Articles

HPTLC separation followed by UV detection of the bitter acid content in various hop samples.

HPTLC separation followed by UV detection of the bitter acid content in various hop samples.

HPTLC separation followed by UV detection of the bitter acid content in various hop samples.

HPTLC separation followed by UV detection of the bitter acid content in various hop samples.

Protocols

HPTLC was used to analyze caffeine in coffee. Filtration of coffee through a PTFE syringe filter was the only sample preparation. Quantitation was conducted with a TLC scanner.

HPTLC was used to analyze caffeine in coffee. Filtration of coffee through a PTFE syringe filter was the only sample preparation. Quantitation was conducted with a TLC scanner.

HPTLC was used to analyze caffeine in coffee. Filtration of coffee through a PTFE syringe filter was the only sample preparation. Quantitation was conducted with a TLC scanner.

HPTLC was used to analyze caffeine in coffee. Filtration of coffee through a PTFE syringe filter was the only sample preparation. Quantitation was conducted with a TLC scanner.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service