If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf
Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
grade
ACS reagent
Quality Segment
product line
EMSURE®
assay
≥99.5% (acidimetric)
form
solid
autoignition temp.
470 °C
potency
2260 mg/kg LD50, oral (Rat)
impurities
≤0.001% Nitrogen compounds (as N), ≤0.5% Water (Karl Fischer), ≤200 ppm Sulfated ash (600 °C), ≤50 ppm In water insoluble matter, ≤5000 ppm Fumaric acid (HPLC)
pH
2.7 (20 °C, 10 g/L in H2O)
bp
235 °C (lit.), 235 °C/1013 hPa
mp
184-186 °C (lit.)
transition temp
flash point 206 °C
solubility
58 g/L
density
1.56 g/cm3 at 20 °C
bulk density
940 kg/m3
anion traces
chloride (Cl-): ≤5 ppm, phosphate (PO43-): ≤0.001%, sulfate (SO42-): ≤30 ppm
cation traces
Fe: ≤5 ppm, heavy metals (as Pb): ≤5 ppm
storage temp.
15-25°C
SMILES string
OC(=O)CCC(O)=O
InChI
1S/C4H6O4/c5-3(6)1-2-4(7)8/h1-2H2,(H,5,6)(H,7,8)
InChI key
KDYFGRWQOYBRFD-UHFFFAOYSA-N
Application
- Anoxic photolysis as a Route to Primary Metabolites on Early Earth: Discusses the transformation of succinic acid in the context of chemical processes on the early Earth, highlighting its potential role in prebiotic chemistry (Taran & Unal, 2024).
- Incorporation of zinc into the protic imidazolium-based ionic liquid: Explores a novel catalytic route using succinic acid for the production of ester plasticizers, highlighting the efficiency of this process in industrial applications (Latos et al., 2024).
Analysis Note
Identity (IR): passes test
Appearance of solution (5 %; water): passes test
In water insoluble matter: ≤ 50 ppm
Melting point: 185.0 - 190.0 °C
Chloride (Cl): ≤ 5 ppm
Phosphate (PO₄): ≤ 0.001 %
Sulfate (SO₄): ≤ 30 ppm
Heavy metals (as lead): ≤ 5 ppm
Fe (Iron): ≤ 5 ppm
Nitrogen compounds (as N): ≤ 0.001 %
Fumaric acid (HPLC): ≤ 5000 ppm
Sulfated ash (600 °C): ≤ 200 ppm
Water (according to Karl Fischer): ≤ 0.5 %
Other Notes
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| grade ACS reagent | grade ACS reagent, puriss. p.a. | grade ACS reagent, anhydrous | grade - |
| assay ≥99.5% (acidimetric) | assay ≥99.5% (T) | assay ≥99.0% | assay 99.0-100.5% (HPLC) |
| Quality Level 300 | Quality Level 200 | Quality Level 200 | Quality Level 500 |
| form solid | form - | form powder | form crystalline, solid |
| storage temp. 15-25°C | storage temp. - | storage temp. - | storage temp. 2-30°C |
| pH 2.7 (20 °C, 10 g/L in H2O) | pH - | pH - | pH - |
Still not finding the right product?
Explore all of our products under Succinic acid
signalword
Danger
hcodes
Hazard Classifications
Eye Dam. 1
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
How can I determine the shelf life / expiration / retest date of this product?
1 answer-
Helpful?
-
-
How is shipping temperature determined? And how is it related to the product storage temperature?
1 answer-
Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf
Helpful?
-


