Skip to Content
Merck
All Photos(4)

Key Documents

797987

Sigma-Aldrich

Succinic acid

anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%

Synonym(s):

Butanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH2CH2COOH
CAS Number:
Molecular Weight:
118.09
Beilstein:
1754069
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
form:
powder
Assay:
≥99.0%
Pricing and availability is not currently available.

grade

ACS reagent
anhydrous

Quality Level

product line

Redi-Dri

Assay

≥99.0%

form

powder

impurities

≤0.001% N compounds
≤0.001% insolubles

ign. residue

≤0.02%

bp

235 °C (lit.)

mp

184-186 °C (lit.)

solubility

methanol: soluble 0.4 g/10 mL

anion traces

chloride (Cl-): ≤0.001%
phosphate (PO43-): ≤0.001%
sulfate (SO42-): ≤0.003%

cation traces

Fe: ≤5 ppm
heavy metals (as Pb): ≤5 ppm

functional group

carboxylic acid

SMILES string

OC(=O)CCC(O)=O

InChI

1S/C4H6O4/c5-3(6)1-2-4(7)8/h1-2H2,(H,5,6)(H,7,8)

InChI key

KDYFGRWQOYBRFD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Succinic acid, also referred to as butanedioic acid, is an organic acid, which can be synthesized by various microorganisms from different carbon sources. Succinic acid has a wide range of applications in agricultural, food and pharmaceutical industries. It is also utilized as a raw material in the industrial chemical synthesis of numerous chemicals including adipic acid, 1,4-butanediol, tetrahydrofuran, N-methyl pyrrolidinone, 2-pyrrolidinone, succinate salts and γ-butyrolactone. In addition, succinic acid can also be used in the synthesis of biodegradable polymers such as polybutyrate succinate (PBS), polyamides and green solvents.[1]

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Production of succinic acid by bacterial fermentation
Hyohak Song a, Sang Yup Lee
Enz. Microbiol. Technol., 39, 352-361 (2006)

Articles

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Redi-Dri™ prevents hygroscopic powders, such as inorganic salts, from absorbing moisture and forming clumps, leaving the salts free-flowing every time.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service