Skip to Content
Merck
All Photos(1)

Documents

05-717

Sigma-Aldrich

Anti-Adenosine Receptor A2a Antibody, clone 7F6-G5-A2

clone 7F6-G5-A2, Upstate®, from mouse

Synonym(s):

adenosine A2 receptor, adenosine A2a receptor, adenosine receptor subtype A2a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

7F6-G5-A2, monoclonal

species reactivity

mouse, rat, dog, canine, rabbit, human

manufacturer/tradename

Upstate®

technique(s)

flow cytometry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ADORA2A(135)

General description

Adenosine receptors, a class of purinergic G-protein coupled receptors with adenosine as endogenous ligands mediate several receptor subtype-specific physiological processes that include cardiac rate, smooth muscle tone, platelet aggregation, inflammation, cell growth and death, and neurotransmission. The activity of A2A adenosine receptor is mediated by G proteins which activate adenylyl cyclase. It is abundant in basal ganglia, vasculature and platelets and it is a major target of caffeine. Adenosine receptor A2A-R contributes to motoneuron survival by transactivating the tyrosine kinase receptor TrkB and may suggest a new strategy for using this mechanism in neurodegenerative diseases.

Specificity

Cross-reactivity with guinea pig also reported.
This antibody recognizes adenosine receptor A2A, Mr 43 kDa.

Immunogen

6His-tagged full-length human recombinant A2A receptor. Epitope mapped to SQPLPGER in the third intracellular loop. Clone 7F6-G5-A2.

Application

Anti-Adenosine Receptor A2a Antibody, clone 7F6-G5-A2 is an antibody against Adenosine Receptor A2a clone 7F6-G5-A2 for use in FC, IH & WB.
Immunohistochemistry: 1 μg/mL of a previous lot detected Adenosine Receptor A2A in paraformaldehyde-fixed mouse brain striatal sections. Not detected in Adenosine Receptor A2A knockout mice.

FACS Analysis: A previous lot of this antibody has been reported by an independent laboratory to detect Adenosine Receptor A2A in transfected HEK293 cells (Koshiba, M., 1999).
Research Category
Neuroscience
Research Sub Category
Neurofilament & Neuron Metabolism

Neuronal & Glial Markers

Neurodegenerative Diseases

Quality

Routinely evaluated by immunohistochemistry.

Western Blot Analysis:
A 1:1000–1:5000 dilution of this lot of antibody detected Adenosine Receptor A2A in HEK293 cells expressing canine A1/A2A receptor chimeras (Rosin, D.L., 1998).

Target description

~43 kDa

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2ak in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt

Analysis Note

Control
Brain tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ultrastructural localization of adenosine A2A receptors suggests multiple cellular sites for modulation of GABAergic neurons in rat striatum.
Hettinger, B D, et al.
The Journal of Comparative Neurology, 431, 331-346 (2001)
Adenosine A2A receptor in the monkey basal ganglia: Ultrastructural localization and colocalization with the metabotropic glutamate receptor 5 in the striatum.
James W Bogenpohl,Stefanie L Ritter,Randy A Hall,Yoland Smith
The Journal of Comparative Neurology null
Deficits in endogenous adenosine formation by ecto-5'-nucleotidase/CD73 impair neuromuscular transmission and immune competence in experimental autoimmune myasthenia gravis.
Oliveira, L; Correia, A; Cristina Costa, A; Guerra-Gomes, S; Ferreirinha et al.
Mediators of Inflammation null
Overexpression of Adenosine A2A Receptors in Rats: Effects on Depression, Locomotion, and Anxiety.
Coelho, JE; Alves, P; Canas, PM; Valadas, JS; Shmidt, T; Batalha, VL; Ferreira et al.
Frontiers in Psychiatry null
Immunohistochemical localization of adenosine A2A receptors in the rat central nervous system
Rosin, D. L., et al
The Journal of Comparative Neurology, 401, 163-186 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service