Skip to Content
Merck
All Photos(5)

Key Documents

05-636

Sigma-Aldrich

Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301

clone JBW301, Upstate®, from mouse

Synonym(s):

H2AXS139P, Histone H2A.X (phospho S139)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

affinity purified immunoglobulin

clone

JBW301, monoclonal

species reactivity

vertebrates

packaging

antibody small pack of 25 μg

manufacturer/tradename

Upstate®

technique(s)

ChIP: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pSer139)

General description

Histone H2A is one of the 5 main histone proteins involved in the structure of chromatin in eukaryotic cells. Featuring a main globular domain and a long N terminal tail H2A is involved with the structure of the nucleosomes of the ′beads on a string′ structure.

Specificity

Anti-phospho-Histone H2A.X (Ser139) Antibody clone JBW301, recognizes Histone H2A.X phosphorylated at Ser139.

Immunogen

peptide (C-KATQA[pS]QEY) corresponding to amino acids 134-142 of human histone H2A.X

Application

Additional Referenced Applications:
Immunohistochemistry Analysis: A representative lot detected Histone H2A.X (pSer139) in RNF168-WT and RNF 168-SA/SEKI mice lung tissue sections (Paraffin). (Xe, X., et al. (2015) Nat. Cell Biol. 20 (3); 320-331).
Chromatin Immunoprecipitation, see Meier, Andreas, et al. EMBO J., 26: 2707-18 (2007) in technical information tab.
Anti-phospho-Histone H2A.X (Ser139), clone JBW301 is a well published Mouse Monoclonal Antibody validated in ChIP, ICC, IF, WB. This purified mAb is highly specific for phospho-Histone H2A.X (Ser139) also known as H2AXS139p.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones

Quality

Immunoblot Analysis: 0.05-1 μg/ml of this antibody detected phosphorylated histone H2A.X (Ser139) in acid extracted histone lysates from Jurkat cells treated with 0.5 μM staurosporine (Catalog # 19-123).
Immunocytochemistry: 2 μg/ml of this antibody detected phosphorylated histone H2A.X in HeLa cells treated with 0.5 μM staurosporine for 4-6 hours.

Target description

17 kDa

Linkage

Replaces: MABE205

Physical form

Format: Purified
Immunoaffinity Purified immunoglobulin in 0.1M Tris-Glycine,pH 7.4, 0.15M NaCl, 0.05% sodium azide as a preservative.
Protein G Purified

Storage and Stability

Maintain for 1 year at 2 to 8°C from date of shipment. For maximum recovery of product, centrifuge the original vial prior to removing the cap.

Analysis Note

Control
UV-treated 293 cell extracts, UV-treated HeLa cell extracts or breast cancer tissue

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of Ku80 deficiency on mutation frequencies and spectra at a LacZ reporter locus in mouse tissues and cells.
Busuttil, RA; Mu?oz, DP; Garcia, AM; Rodier, F; Kim, WH; Suh, Y; Hasty, P; Campisi, J; Vijg, J
Testing null
Unsuitability of lymphoblastoid cell lines as surrogate of cryopreserved isolated lymphocytes for the analysis of DNA double-strand break repair activity.
Zijno, Andrea, et al.
Mutation Research, 684, 98-105 (2010)
FEN1 contributes to telomere stability in ALT-positive tumor cells.
Saharia, A; Stewart, SA
Oncogene null
Irene V Bijnsdorp et al.
Cancer science, 101(2), 440-447 (2009-11-06)
The pyrimidine trifluorothymidine (TFT) inhibits thymidylate synthase (TS) and can be incorporated into the DNA. TFT, as part of TAS-102, is clinically evaluated in phase II studies as an oral chemotherapeutic agent. Erlotinib is a tyrosine kinase inhibitor of the
Lidia Ruiz et al.
PloS one, 3(9), e3230-e3230 (2008-09-19)
P53 activation can trigger various outcomes, among them reversible growth arrest or cellular senescence. It is a live debate whether these outcomes are influenced by quantitative or qualitative mechanisms. Furthermore, the relative contribution of p53 to Ras-induced senescence is also

Articles

Explore the basics of working with antibodies including technical information on structure, classes, and normal immunoglobulin ranges.

Explore the basics of working with antibodies including technical information on structure, classes, and normal immunoglobulin ranges.

Antibodies combine with specific antigens to generate an exclusive antibody-antigen complex. Learn about the nature of this bond and its use as a molecular tag for research.

Antibodies combine with specific antigens to generate an exclusive antibody-antigen complex. Learn about the nature of this bond and its use as a molecular tag for research.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service