Skip to Content
Merck
All Photos(2)

Key Documents

198269

Sigma-Aldrich

Erythrosin B

certified by the Biological Stain Commission, Dye content ≥85 %

Synonym(s):

Erythrosin extra bluish, 2′,4′,5′,7′-Tetraiodofluorescein disodium salt, Acid Red 51, Iodoeosin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H6I4Na2O5
CAS Number:
Molecular Weight:
879.86
Colour Index Number:
45430
Beilstein:
1443945
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

certified by the Biological Stain Commission

Quality Level

form

powder

composition

Dye content, ≥85%

technique(s)

microbe id | staining: suitable

color

dark red

pH

6.4 (30.3 °C)

mp

315 °C

solubility

H2O: 1 mg/mL, clear

density

0.625 — 0.731 g/cm3 at 30.1 °C (86.2 °F)

λmax

525 nm

ε (extinction coefficient)

≥13000 at 308-312 nm
≥32000 at 259-263 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].[Na+].[O-]c1c(I)cc2c(Oc3c(I)c([O-])c(I)cc3C24OC(=O)c5ccccc45)c1I

InChI

1S/C20H8I4O5.2Na/c21-11-5-9-17(13(23)15(11)25)28-18-10(6-12(22)16(26)14(18)24)20(9)8-4-2-1-3-7(8)19(27)29-20;;/h1-6,25-26H;;/q;2*+1/p-2

InChI key

RAGZEDHHTPQLAI-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Erythrosin B has been used as a dye in fluorescence lifetime imaging.

Biochem/physiol Actions

Erythrosin B, also called as eosin B, is a xanthene dye. It belongs to the family of fluorescein dyes. Erythrosin B is an artificial food dye that induces hyperkinesis when swallowed by susceptible children. It plays a major role in inhibiting dopamine uptake and high affinity 3H-ouabain binding and ion transport in synaptosomes from rat caudate nucleus.

Suitability

Counterstain to alum-hematoxylin.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

359.6 °F - closed cup

Flash Point(C)

182 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erythrosin B is a specific inhibitor of high affinity 3H-ouabain binding and ion transport in rat brain
Silbergeld E K
Neuropharmacology, 20:1, 87-90 (1981)
C. R. Acad. Sci. (D), 179, 367-367 (1924)
P Johnson et al.
The Biochemical journal, 203(1), 313-321 (1982-04-01)
We have previously described a method for measuring the rotational diffusion of membrane proteins by using fluorescent triplet probes [Johnson & Garland (1981) FEBS Lett. 135, 252-256]. We now describe the criteria by which the suitability of such probes may
CD40-targeting KGYY15 peptides do not efficiently block the CD40-CD40L interaction.
Damir Bojadzic et al.
Diabetologia, 62(11), 2158-2160 (2019-09-11)
Bright monomeric red fluorescent protein with an extended
fluorescence lifetime
Merzlyak EM
Nature Methods, 4(7), 555-557 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service