Skip to Content
Merck
All Photos(1)

Documents

05-577

Sigma-Aldrich

Anti-PP2A Antibody, C subunit, demethylated, clone 4b7

ascites fluid, clone 4b7, Upstate®

Synonym(s):

Anti-NEDLBA, Anti-PP2Ac, Anti-PP2CA, Anti-PP2Calpha, Anti-RP-C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

ascites fluid

antibody product type

primary antibodies

clone

4b7, monoclonal

species reactivity

vertebrates

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PPP2CA(5515)

Related Categories

Specificity

unmethylated PP2A, C subunit

Immunogen

peptide (C-RGEPHVTRRTPDYFL) corresponding to amino acids 295-309 of human PP2A, C subunit

Application

Anti-PP2A Antibody, C subunit, demethylated, clone 4b7 is an antibody against PP2A for use in IC, IP & WB.
Research Category
Signaling

Neuroscience
Research Sub Category
Kinases & Phosphatases

Neurodegenerative Diseases

Quality

routinely evaluated by immunoblot on RIPA lysates from A431 cells

Target description

36kDa

Physical form

0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Ascites

Storage and Stability

2 years at -20°C

Analysis Note

Control
Positive Antigen Control: Catalog #12-301, non-stimulated A431 cell lysate. Add 2.5µL of 2-mercaptoethanol/100µL of lysate and boil for 5 minutes to reduce the preparation. Load 20µg of reduced lysate per lane for minigels.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoyu Xia et al.
Cell cycle (Georgetown, Tex.), 14(12), 1938-1947 (2015-04-04)
Leucine carboxyl methyltransferase-1 (LCMT1) and protein phosphatase methylesterase-1 (PME-1) are essential enzymes that regulate the methylation of the protein phosphatase 2A catalytic subunit (PP2AC). LCMT1 and PME-1 have been linked to the regulation of cell growth and proliferation, but the
Juyeon Hwang et al.
The Journal of biological chemistry, 291(40), 21008-21019 (2016-08-11)
The protein phosphatase 2A (PP2A) subfamily of phosphatases, PP2A, PP4, and PP6, are multifunctional serine/threonine protein phosphatases involved in many cellular processes. Carboxyl methylation of the PP2A catalytic subunit (PP2Ac) C-terminal leucine is regulated by the opposing activities of leucine
Ying Zhou et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 34(12), 16414-16431 (2020-10-19)
Polyphyllin I (PPI) is a natural phytochemical drug isolated from plants which can inhibit the proliferation of cancer cells. One of the PPI tumor-inhibitory effects is through downregulating the expression of Cancerous Inhibitor of PP2A (CIP2A), the latter, is found
Vitali Stanevich et al.
Molecular cell, 41(3), 331-342 (2011-02-05)
Proper formation of protein phosphatase 2A (PP2A) holoenzymes is essential for the fitness of all eukaryotic cells. Carboxyl methylation of the PP2A catalytic subunit plays a critical role in regulating holoenzyme assembly; methylation is catalyzed by PP2A-specific methyltransferase LCMT-1, an
Christina Ising et al.
Nature, 575(7784), 669-673 (2019-11-22)
Alzheimer's disease is characterized by the accumulation of amyloid-beta in plaques, aggregation of hyperphosphorylated tau in neurofibrillary tangles and neuroinflammation, together resulting in neurodegeneration and cognitive decline1. The NLRP3 inflammasome assembles inside of microglia on activation, leading to increased cleavage

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service