Skip to Content
Merck
All Photos(4)

Key Documents

05-421

Sigma-Aldrich

Anti-PP2A Antibody, C subunit, clone 1D6

clone 1D6, Upstate®, from mouse

Synonym(s):

Replication protein C, protein phosphatase 2 (formerly 2A), catalytic subunit, alpha isoform, protein phosphatase 2, catalytic subunit, alpha isoform, serine/threonine protein phosphatase 2A, catalytic subunit, alpha isoform

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1D6, monoclonal

species reactivity

Saccharomyces cerevisiae, bovine, rabbit, rat, Xenopus, mouse, human

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... PPP2CA(5515)

General description

Type 2 protein serine/threonine phosphatase (PP2A) is a trimer composed of a catalytic C subunit and two regulatory (A and B) subunits; many isoforms for the A and B subunits have been found. PP2A has a high specific activity against the α subunit of phosphorylase kinase and is insensitive to Inhibitor-2. PP2A is distinguishable from PP2B/Calcineurin and PP2C by its partial activity in the absence of divalent cations, and by its high sensitivity to inhibition by okadaic acid. It is also thought to be the principal downmodulator of serine/threonine kinase cascades. PP2B requires Ca2+/calmodulin for activity, and is an activator of NFAT transcription factors. PP2C requires Mg2+, but is insensitive to okadaic acid, microcystin LR, or the calmodulin inhibitor trifluoperazine.

Specificity

This antibody recognizes PP2A catalytic subunit, Mr 36 kDa.

Immunogen

A 16 residue synthetic peptide (C-RGEPHVTRRTPDYFL) corresponding to amino acids 295-309 of the 36 kDa catalytic subunit of human protein phosphatase 2A (PP2A). Clone 1D6.
Epitope: a.a. 295-309

Application

Detect PP2A using this Anti-PP2A Antibody, C subunit, clone 1D6 validated for use in IC, IP & WB.
Immunoprecipitation:
4 µg of a previous lot immunoprecipitated PP2A from 500 µg of A431 RIPA lysate. Note: This antibody has been reported not to block phosphatase activity.

Immunocytochemistry:
5-10 µg/mL of a previous lot of this antibody showed positive immunostaining for PP2A in A431 cells fixed for 10 minutes with 4% paraformaldehyde in PBS.
Research Category
Signaling

Neuroscience
Research Sub Category
Neurotransmitters & Receptors

Neurodegenerative Diseases

Quality

Routinely evaluated by western blot on human A431 or murine 3T3/NIH RIPA cell lysates.

Western Blot Analysis:
0.5-2 µg/mL of this lot detected PP2A in human A431 and previously in murine 3T3 RIPA cell lysates.

Target description

36 kDa

Physical form

Format: Purified
Protein A chromatography
Purified mouse monoclonal IgG2bκ in buffer containing 0.1M Tris-Glycine, pH 7.4, 0.15 M, NaCl, and 0.05% sodium azide. Frozen solution.

Storage and Stability

Stable for 1 year at -20ºC from date of receipt. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Analysis Note

Control
Positive Antigen Control: Catalog #12-301, non-stimulated A431 cell lysate. Add 2.5µL of 2-mercaptoethanol/100µL of lysate and boil for 5 minutes to reduce the preparation. Load 20µg of reduced lysate per lane for minigels.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Circumventing embryonic lethality with Lcmt1 deficiency: generation of hypomorphic Lcmt1 mice with reduced protein phosphatase 2A methyltransferase expression and defects in insulin signaling.
MacKay, KB; Tu, Y; Young, SG; Clarke, SG
Testing null
H Wei et al.
The Journal of biological chemistry, 276(2), 1570-1577 (2000-10-20)
Protein phosphatase 2A (PP2A) is an essential eukaryotic serine/threonine phosphatase known to play important roles in cell cycle regulation. Association of different B-type targeting subunits with the heterodimeric core (A/C) enzyme is known to be an important mechanism of regulating
Characterization of the PP2A alpha gene mutation in okadaic acid-resistant variants of CHO-K1 cells
Shima, H, et al
Proceedings of the National Academy of Sciences of the USA, 91, 9267-9271 (1994)
Identification of protein phosphatase 1 as a mitotic lamin phosphatase
Thompson, L. J., et al
The Journal of Biological Chemistry, 272, 29693-29697 (1997)
Generation of Ppp2Ca and Ppp2Cb conditional null alleles in mouse.
Pengyu Gu,Xin Qi,Yue Zhou,Yun Wang,Xiang Gao
Genesis (2000)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service