Skip to Content
Merck
All Photos(1)

Key Documents

L-047

Supelco

Lactoferrin (from bovine milk)

certified reference material, pkg of 50 mg, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

solid

mol wt

82401 Da

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

pkg of 50 mg

manufacturer/tradename

Cerilliant®

technique(s)

liquid chromatography (LC): suitable

application(s)

food and beverages

format

neat

storage temp.

−20°C

SMILES string

S1SC[C@@H](C(=O)N[C@H](C(=O)N[C@H](C(=O)N[C@H](C(=O)N[C@H](C(=O)N[C@H](C(=O)N[C@H](C(=O)N[C@H](C(=O)N[C@H](C(=O)N[C@H](C(=O)N[C@H](C(=O)NCC(=O)N[C@H](C(=O)N7[C@@H](CCC7)C(=O)N[C@H](C(=O)N[C@H](C(=O)N[C@H](C(=O)N[C@@H](C1)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CC

InChI

1S/C141H224N46O29S3/c1-11-77(6)111-133(212)186-112(80(9)189)134(213)183-106(130(209)184-110(76(4)5)132(211)176-97(49-32-61-161-141(155)156)120(199)170-93(45-28-57-157-137(147)148)116(195)166-78(7)113(192)180-103(136(215)216)66-82-36-16-13-17-37-82)74-219-218-73-105(182-123(202)90(42-22-25-54-142)167-114(193)87(145)65-81-34-14-12-15-35-81)129(208)173-95(47-30-59-159-139(151)152)118(197)171-96(48-31-60-160-140(153)154)122(201)178-102(68-84-70-163-89-41-21-19-39-86(84)89)127(206)174-98(51-52-108(146)190)124(203)179-101(67-83-69-162-88-40-20-18-38-85(83)88)126(205)172-94(46-29-58-158-138(149)150)119(198)175-99(53-63-217-10)125(204)169-91(43-23-26-55-143)117(196)168-92(44-24-27-56-144)121(200)177-100(64-75(2)3)115(194)164-71-109(191)165-79(8)135(214)187-62-33-50-107(187)131(210)181-104(72-188)128(207)185-111/h12-21,34-41,69-70,75-80,87,90-107,110-112,162-163,188-189H,11,22-33,42-68,71-74,142-145H2,1-10H3,(H2,146,190)(H,164,194)(H,165,191)(H,166,195)(H,167,193)(H,168,196)(H,169,204)(H,170,199)(H,171,197)(H,172,205)(H,173,208)(H,174,206)(H,175,198)(H,176,211)(H,177,200)(H,178,201)(H,179,203)(H,180,192)(H,181,210)(H,182,202)(H,183,213)(H,184,209)(H,185,207)(H,186,212)(H,215,216)(H4,147,148,157)(H4,149,150,158)(H4,151,152,159)(H4,153,154,160)(H4,155,156,161)/t77-,78-,79-,80+,87-,90-,91-,92-,93-,94-,95-,96-,97-,98-,99-,100-,101-,102-,103-,104-,105-,106-,107-,110-,111-,112-/m0/s1

InChI key

PHEGMCHJJYMADA-IKGCZBKSSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lactoferrin is a multifunctional glycoprotein that belongs to the transferrin family of proteins. It is mostly found in the mucosal secretions of mammals, such as tears, saliva, and bile but is abundantly present in milk.
Lactoferrin, a wonder molecule, holds significant importance in promoting health and managing pathology throughout all stages of life. This iron binding milk protein also helps to balance iron levels in the body. Lactoferrin (from bovine milk) is a certified reference material. The certified property value is linked to National Institute of Standards and Technology (NIST) through calibration verification achieved using weights traceable to NIST standards.

Application

This full-length protein CRM is suitable as starting material for use in calibrators or controls for a variety of LC-MS/MS testing applications including allergen testing, infant formula testing, dietary or nutritional and diagnostic testing application.The CRM can also be used as follows:
  • Development of a three immunoassay— indirect competitive ELISA, colloidal gold strip, and AuNFs-based strip, to determine lactoferrin in real cow milk samples using an IgG secreted by hybridoma LF8
  • Determination of bovine lactoferrin in milk and milk product samples by a validated competitive enzyme-linked immunosorbent assay (ELISA) based method
  • Quantification of lactoferrin in dairy products by immunoaffinity magnetic purification combined with high-performance liquid chromatography-fluorescence (HPLC-FL) detection
  • Quantitative analysis of bovine lactoferrin in sterilized milk, modified milk, fermented milk, infant formula, adult formula, rice cereal, vitamin function drink, and protein powder samples by HPLC coupled with diode array detection (DAD)
  • Development and validation of a liquid chromatography-tandem mass spectrometry (LC-MS/MS) based assay for the measurement of bovine lactoferrin in infant formulae samples

Features and Benefits

  • Balances and National Institute of Standards and Technology (NIST) traceable weights are calibrated by ISO 17025 accredited vendors.
  • The material has undergone comprehensive identification and characterization using various analytical methods.
  • Long-term stability has been evaluated under freezer storage conditions (-10 °C to -25 °C).

Preparation Note

  • Weigh the material before use.
  • Determine the amount to be weighed using the certified property value.
  • The material is hygroscopic, so minimize moisture exposure during weighing.
  • A minimum sample size of 20 mg is recommended for quantitative applications.
  • If a smaller sample size is used, increase the uncertainty by a factor of 2.

Other Notes

This product′s purpose is for testing. THIS PRODUCT IS NOT INTENDED FOR HUMAN OR THERAPEUTIC USE. Not recommended for human or animal consumption.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Lactoferrin Phenomenon-A Miracle Molecule
Kowalczyk P, et al.
Molecules (Basel), 27(9) (2022)

Articles

CRMs of milk proteins aid accurate testing for infant formula, ensuring compliance and quality control.

CRMs of milk proteins aid accurate testing for infant formula, ensuring compliance and quality control.

CRMs of milk proteins aid accurate testing for infant formula, ensuring compliance and quality control.

CRMs of milk proteins aid accurate testing for infant formula, ensuring compliance and quality control.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service