Skip to Content
Merck

L3770

Lactoferrin from human milk

Iron saturated, ≥85% (SDS-PAGE)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352202
NACRES:
NA.61
MDL number:
Form:
powder
Assay:
≥85% (SDS-PAGE)
Biological source:
human milk
Mol wt:
82.4 kDa
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human milk

Quality Segment

assay

≥85% (SDS-PAGE)

form

powder

mol wt

82.4 kDa

Iron content

0.15-1.0%

technique(s)

microbiological culture: suitable

solubility

phosphate buffer: 10 mg/mL at 25 °C (0.01 M phosphate buffer, 0.0027 M potassium chloride and 0.137 M sodium chloride, pH 7.4)

UniProt accession no.

storage temp.

2-8°C

SMILES string

N([C@@H](C(C)C)C(=O)N[C@@H](C)C(=O)N[C@@H](Cc1ccccc1)C(=O)N[C@@H](C(C)C)C(=O)O)C(=O)[C@@H](NC(=O)CNC(=O)[C@@H](NC(=O)[C@@H](N)CC(=O)N)C)CC(=O)O

InChI

1S/C35H53N9O12/c1-16(2)27(43-33(53)23(14-26(47)48)41-25(46)15-38-29(49)18(5)39-31(51)21(36)13-24(37)45)34(54)40-19(6)30(50)42-22(12-20-10-8-7-9-11-20)32(52)44-28(17(3)4)35(55)56/h7-11,16-19,21-23,27-28H,12-15,36H2,1-6H3,(H2,37,45)(H,38,49)(H,39,51)(H,40,54)(H,41,46)(H,42,50)(H,43,53)(H,44,52)(H,47,48)(H,55,56)/t18-,19-,21-,22-,23-,27-,28-/m0/s1

InChI key

QCBUWCQOKPLTDZ-PKRULZLPSA-N

Gene Information

human ... LTF(4057)

Application

Lactoferrin was used to grow Streptococcus mutans in an iron-limiting medium. It was used to test if lactoferrin impedes epithelial cell adhesion in vitro. It was also used to test the diagnostic value of blood cytokine concentrations in acute pneumonia.

Biochem/physiol Actions

Lactoferrin is an iron binding protein. It is structurally similar to transferrin, the plasma iron transport protein; but lactoferrin has a much higher affinity for iron (250 fold). It is very abundant in colostrum and small amounts can also be found in tears, saliva, mucous secretions and in the secondary granules of neutrophils. It is made by mucosal epithelium and neutrophils and is released by these cells in response to inflammatory stimuli. Bacterial growth is inhibited by its ability to sequester iron and also permeabilize bacterial cell walls by binding to lipopolysaccharides through its N-terminus. Lactoferrin can inhibit viral infection by binding tightly to the viral envelope protein. This prevents cell-virus fusion by blocking the binding domain. Lactoferrin appears to activate host defense systems in part by stimulating the release of interleukin-8, a neutrophil activator. It may also be involved in antibody and interleukin synthesis, lymphocyte proliferation and complement activation.


Still not finding the right product?

Explore all of our products under Lactoferrin from human milk




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library