Skip to Content
Merck
All Photos(1)

Key Documents

H-001

Supelco

Hydrochlorothiazide solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8ClN3O4S2
CAS Number:
Molecular Weight:
297.74
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

NS(=O)(=O)c1cc2c(NCNS2(=O)=O)cc1Cl

InChI

1S/C7H8ClN3O4S2/c8-4-1-5-7(2-6(4)16(9,12)13)17(14,15)11-3-10-5/h1-2,10-11H,3H2,(H2,9,12,13)

InChI key

JZUFKLXOESDKRF-UHFFFAOYSA-N

Gene Information

human ... SLC12A3(6559)

General description

Hydrochlorothiazide is a calcium-sparing diuretic used for treatment of edema and hypertension. A thiazide derivative, hydrochlorothiazide is sold under the trade names Microzide® and Aquazide H. This Certified Spiking Solution® is suitable for use in LC/MS or GC/MS applications such as clinical toxicology and forensic analysis.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Microzide is a registered trademark of Watson Laboratories, Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Successful rapid desensitization to hydrochlorothiazide.
Jamma Li et al.
Annals of allergy, asthma & immunology : official publication of the American College of Allergy, Asthma, & Immunology, 110(4), 307-308 (2013-03-29)
Yoshihisa Hanayama et al.
Acta medica Okayama, 66(6), 449-459 (2012-12-21)
Angiotensin receptor blockers (ARBs) are the first-line antihypertensive agents. In clinical practice, it is often difficult to achieve the recommended blood pressure level by ARBs in their ordinal dosages alone. This study examined the practical efficacy of a combination therapy
Alexander V Murray et al.
Journal of clinical hypertension (Greenwich, Conn.), 14(12), 821-827 (2012-12-05)
Combination antihypertensive therapies are recommended to attain blood pressure (BP) targets especially in high-risk patients in whom rapid and pronounced BP control is essential. This 28- to 54-week, open-label, multicenter study evaluated the safety and efficacy of a triple combination
Michael A Weber et al.
Lancet (London, England), 381(9866), 537-545 (2012-12-12)
In previous clinical trials in high-risk hypertensive patients, paradoxically higher cardiovascular event rates have been reported in patients of normal weight compared with obese individuals. As a prespecified analysis of the Avoiding Cardiovascular Events through Combination Therapy in Patients Living
Krzysztof Łukawski et al.
Acta poloniae pharmaceutica, 70(1), 147-152 (2013-04-25)
Research data show that diuretics can have anticonvulsant properties. This study examined effects of ethacrynic acid, a loop diuretic, and hydrochlorothiazide, a thiazide-type diuretic, on the anticonvulsant activity of gabapentin, a newer antiepileptic drug, in the maximal electroshock seizure threshold

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service