Skip to Content
Merck
All Photos(1)

Documents

CDS021593

Sigma-Aldrich

Pioglitazone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H20N2O3S
Molecular Weight:
356.44
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

description

AldrichCPR

form

solid

SMILES string

O=C(N1)C(SC1=O)CC(C=C2)=CC=C2OCCC(N=C3)=CC=C3CC

InChI

1S/C19H20N2O3S/c1-2-13-3-6-15(20-12-13)9-10-24-16-7-4-14(5-8-16)11-17-18(22)21-19(23)25-17/h3-8,12,17H,2,9-11H2,1H3,(H,21,22,23)

InChI key

HYAFETHFCAUJAY-UHFFFAOYSA-N

Gene Information

human ... PPARG(5468)

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH′S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT "AS-IS" AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY, (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE, OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY, WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lourdes Ibáñez et al.
The Journal of adolescent health : official publication of the Society for Adolescent Medicine, 61(4), 446-453 (2017-07-18)
Polycystic ovary syndrome (PCOS) is an increasingly prevalent disorder in adolescent girls, commonly presenting with hirsutism/oligomenorrhea, commonly treated with an oral contraceptive (OC), and commonly followed by oligoanovulatory subfertility. We tested whether an intervention targeting the reduction of hepato-visceral adiposity
Tsung-Yi Cheng et al.
Journal of the Chinese Medical Association : JCMA, 80(11), 683-689 (2017-10-04)
Hepatopulmonary syndrome (HPS) is characterized by oxygen desaturation and increased intrapulmonary shunting formation in cirrhosis. Due to an unclarified mechanism, there is still no effective therapy except liver transplantation. Recent studies revealed that pulmonary angiogenesis may participate in pathogenesis, in
Latifa Bakiri et al.
The Journal of experimental medicine, 214(5), 1387-1409 (2017-03-31)
Human hepatocellular carcinomas (HCCs), which arise on a background of chronic liver damage and inflammation, express c-Fos, a component of the AP-1 transcription factor. Using mouse models, we show that hepatocyte-specific deletion of c-Fos protects against diethylnitrosamine (DEN)-induced HCCs, whereas
Joshua D Rosenblat
Psychopharmacology, 236(10), 2909-2921 (2019-02-14)
Immune dysfunction has been strongly implicated in the pathophysiology of bipolar disorder (BD). As such, numerous clinical trials have investigated the effects of anti-inflammatory agents in the treatment of BD. Review clinical studies evaluating the effects of anti-inflammatory agents in
Qing-Zhang Tuo et al.
Signal transduction and targeted therapy, 7(1), 59-59 (2022-02-25)
Ischemic stroke represents a significant danger to human beings, especially the elderly. Interventions are only available to remove the clot, and the mechanism of neuronal death during ischemic stroke is still in debate. Ferroptosis is increasingly appreciated as a mechanism

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service