Skip to Content
Merck
All Photos(1)

Key Documents

F-043

Supelco

Flubromazepam

1.0 mg/mL in methanol, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10BrFN2O
CAS Number:
Molecular Weight:
333.16
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

−20°C

SMILES string

O=C1CN=C(C2=CC=CC=C2F)C3=C(C=CC(Br)=C3)N1

InChI

1S/C15H10BrFN2O/c16-9-5-6-13-11(7-9)15(18-8-14(20)19-13)10-3-1-2-4-12(10)17/h1-7H,8H2,(H,19,20)

InChI key

ZRKDDZBVSZLOFS-UHFFFAOYSA-N

General description

Flubromazepam is a potent, long-acting novel psychoactive benzodiazepine with anxiolytic, anticonvulsant, muscle relaxant and hypnotic properties. Flubromazepam reportedly has a high addiction potential and risk of overdose. This certified Snap-N-Spike® solution is suitable for use as a starting material in calibrators and controls for numerous GC/MS or LC/MS applications in forensic analysis, clinical toxicology and urine drug testing.

Application


  • Quantitative analysis of benzodiazepines: Flubromazepam was included in a validated LC-MS/MS method for quantifying 13 designer benzodiazepines in blood, highlighting its importance in forensic toxicology and psychoactive substance studies (Mei et al., 2019).

  • Retrospective HRMS data application: Flubromazepam′s detection in complex drug mixtures via high-resolution mass spectrometry underscores its application in enhancing analytical methodologies for drug safety and pharmacological research (Partridge et al., 2018).

  • Advanced chromatographic techniques: The compound was part of a study using ultra-high performance liquid chromatography coupled with high-resolution time-of-flight mass spectrometry, improving the precision in detecting benzodiazepine analogs within pharmaceutical and clinical research contexts (Tomková et al., 2017).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service