Skip to Content
Merck
All Photos(1)

Key Documents

A-916

Supelco

7-Aminoclonazepam solution

1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12ClN3O
CAS Number:
Molecular Weight:
285.73
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

SNAP-N-SPIKE®, SNAP-N-SHOOT®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in acetonitrile

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

2-8°C

SMILES string

NC1=CC=C2C(C(C3=C(Cl)C=CC=C3)=NCC(N2)=O)=C1

InChI

1S/C15H12ClN3O/c16-12-4-2-1-3-10(12)15-11-7-9(17)5-6-13(11)19-14(20)8-18-15/h1-7H,8,17H2,(H,19,20)

InChI key

HEFRPWRJTGLSSV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified Snap-N-Spike® solution is suitable for use as a starting material in calibrators and controls for numerous GC/MS or LC/MS applications in forensic analysis, clinical toxicology and urine drug testing. As a major metabolite, 7-aminoclonazepam may be used to monitor use of the parent drug, clonazepam. Clonazepam, marketed as Klonopin® and Rivotril, is a long-acting benzodiazepine with anxiolytic, anticonvulsant, muscle relaxant, and hypnotic properties.

Application


  • 7-Aminoclonazepam as a Reference Standard in Forensic Toxicology: This benzodiazepine metabolite is critical for forensic applications, where it serves as a reference standard for the confirmation and quantitation of clonazepam in biological samples. Its reliable detection in toxicological screenings is crucial for legal and medical interpretations (Elenstål et al., 2023).

  • Pharmacokinetic Studies with 7-Aminoclonazepam: In pharmacokinetics, 7-Aminoclonazepam is studied to understand the metabolic pathways of clonazepam, particularly its transformation and elimination in the human body. This research is vital for developing dosing regimens that optimize therapeutic outcomes while minimizing side effects (Øiestad et al., 2023).

  • Stability Studies of 7-Aminoclonazepam: Stability testing of 7-Aminoclonazepam in various storage conditions provides essential data on how storage conditions affect the integrity of this compound. Such information ensures the accuracy and reliability of bioanalytical assays used in clinical and forensic settings (Dahlin & Petrides, 2022).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Klonopin is a registered trademark of Hoffmann-La Roche Inc.
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

35.6 °F - closed cup

Flash Point(C)

2 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simple high-performance liquid chromatographic assay for the routine monitoring of clonazepam in plasma.
R L Heazlewood et al.
Journal of chromatography, 336(1), 229-233 (1984-12-07)
Wei Chen et al.
Biosensors & bioelectronics, 24(7), 2051-2056 (2008-12-09)
The present paper described a rapid and ultrasensitive detection method using a microfluidic chip for analyzing 7-aminoclonazepam (7-ACZP) residues in human urine. A microfluidic chip-based immunoassay with laser-induced fluorescence (LIF) detection based on the water-soluble denatured bovine serum albumin (dBSA)-coated
D R Peng et al.
Pediatric pharmacology (New York, N.Y.), 4(3), 155-159 (1984-01-01)
The N-acetylation of the reduced metabolite of clonazepam 7-amino-clonazepam was studied in cytosolic preparation from human fetal and adult livers. The metabolite formed 7-acetamido-clonazepam was measured with high performance liquid chromatography. A bimodal distribution of the N-acetyltransferase activities was observed
A Negrusz et al.
Journal of analytical toxicology, 24(7), 614-620 (2000-10-24)
Clonazepam (CLO) is an anticonvulsant benzodiazepine approved by the Food and Drug Administration for use in the treatment of seizures. It produces pharmacological effects (depression, amnesia) similar to other compounds from the same therapeutic class, and in combination with alcohol
Albert A Elian
Forensic science international, 134(1), 54-56 (2003-07-05)
The ability of five commercially available enzyme-linked immunosorbent assay (ELISA) benzodiazepines to detect clonazepam and 7-aminoclonazepam in blood and urine was investigated. To determine the cross-reactivity of various ELISA assays, drug free blood and urine were fortified with clonazepam and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service