Skip to Content
Merck
All Photos(1)

Documents

T-911

Supelco

α-Hydroxytriazolam solution

1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H12Cl2N4O
CAS Number:
Molecular Weight:
359.21
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

form

liquid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

concentration

1.0 mg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

clinical testing

format

single component solution

storage temp.

2-8°C

SMILES string

OCc1nnc2CN=C(c3ccccc3Cl)c4cc(Cl)ccc4-n12

InChI

1S/C17H12Cl2N4O/c18-10-5-6-14-12(7-10)17(11-3-1-2-4-13(11)19)20-8-15-21-22-16(9-24)23(14)15/h1-7,24H,8-9H2

InChI key

BHUYWUDMVCLHND-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

a-Hydroxytriazolam is a major urinary and blood metabolite of the benzodiazepine, triazolam, a pharmaceutical used to treat severe insomnia. This certified solution standard is suitable for use in LC/MS or GC/MS applications for clinical toxicology, forensic analysis, urine drug testing, or pharmaceutical research.

Application


  • Pharmacokinetic analysis of benzodiazepines: alpha-Hydroxytriazolam solution is employed in the quantitation of benzodiazepines and their metabolites in biological fluids using liquid chromatography-tandem mass spectrometry, providing accurate pharmacokinetic profiles that are essential for clinical and forensic analysis (Zheng et al., 2018).

  • Receptor binding studies: The application of high-purity alpha-Hydroxytriazolam in receptor binding studies enables researchers to elucidate the interaction mechanisms of benzodiazepine analogs at the molecular level, thereby enhancing the understanding of their pharmacological effects (Minegishi et al., 2021).

  • Development of analytical methods: alpha-Hydroxytriazolam solution serves as a critical analytical standard in the development of robust assays for the detection and quantification of benzodiazepines in various matrices, facilitating advancements in toxicological and pharmacological research (Lahr et al., 2020).

  • Forensic and clinical toxicology: Utilized in forensic science, alpha-Hydroxytriazolam solution aids in the rapid determination of benzodiazepines and their metabolites in urine samples, crucial for the accurate interpretation of toxicological data (Jeong et al., 2015).

  • Scientific research in biochemistry: alpha-Hydroxytriazolam research compounds are vital for studying the metabolic pathways and degradation profiles of triazolam and related benzodiazepines, supporting biochemists in drug metabolism and pharmacokinetics research (Jin et al., 2011).

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F - closed cup

Flash Point(C)

9.7 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

R L O'Connor-Semmes et al.
Clinical pharmacology and therapeutics, 70(2), 126-131 (2001-08-15)
This study evaluated the effect of oral ranitidine (75 mg and 150 mg) on the pharmacokinetics of triazolam (0.25 mg) and its major metabolite, alpha-hydroxytriazolam, in both young and older people. Metabolite data were used to distinguish the mechanism of
Kenji Tsujikawa et al.
Journal of analytical toxicology, 29(4), 240-243 (2005-06-25)
The objective of this study was to examine urinary excretion profiles of two major triazolam metabolites, alpha-hydroxytriazolam (alpha-OHTRZ) and 4-hydroxytriazolam (4-OHTRZ) in humans. Urine samples were collected from three healthy male volunteers who had been previously administered single 0.25- and
F S Eberts et al.
Clinical pharmacology and therapeutics, 29(1), 81-93 (1981-01-01)
Triazolam (T) is a new, potent hypnotic with a short duration of action in man. After an 0.88-mg oral dose of T-14C in six male subjects, mean recovery of 14C radioactivity was 85% in urine and 8% in feces. The
A D Fraser et al.
Journal of analytical toxicology, 16(6), 347-350 (1992-11-01)
Triazolam is a very short-acting triazolobenzodiazepine with sedative-hypnotic properties. Approximately 2% of an oral dose is excreted unchanged in the urine. The major urinary metabolite is alpha-hydroxytriazolam glucuronide (70% of the dose). The objective of this study was to characterize
Dora Farkas et al.
Journal of clinical pharmacology, 47(3), 286-294 (2007-02-27)
The effect of pomegranate juice (PJ) or grapefruit juice (GFJ) on CYP3A activity was studied in vitro and in healthy human volunteers. In human liver microsomes, the mean 50% inhibitory concentrations (IC(50)) for PJ and GFJ versus CYP3A (triazolam alpha-hydroxylation)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service