Skip to Content
Merck
All Photos(1)

Key Documents

999992P

Avanti

16:0 Diether PC

Avanti Research - A Croda Brand 999992P, powder

Synonym(s):

1,2-di-O-hexadecyl-sn-glycero-3-phosphocholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H84NO6P
CAS Number:
Molecular Weight:
706.07
MDL number:
UNSPSC Code:
51191904
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 10 mg (999992P-10mg)

manufacturer/tradename

Avanti Research - A Croda Brand 999992P

lipid type

phospholipids
cardiolipins

shipped in

dry ice

storage temp.

−20°C

SMILES string

[O-]P(OCC[N+](C)(C)C)(OC[C@]([H])(OCCCCCCCCCCCCCCCC)COCCCCCCCCCCCCCCCC)=O

InChI

1S/C40H84NO6P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-35-44-38-40(39-47-48(42,43)46-37-34-41(3,4)5)45-36-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h40H,6-39H2,1-5H3

InChI key

MWTIGLPPQBNUFP-UHFFFAOYSA-N

General description

16:0 Diether PC (phosphocholine) is a zwitterionic lipid having choline as the alcohol moiety, attached to the phosphate group. It has two hexadecane chains attached to the sn-1 and sn-2 positions of the glycerol backbone by ether bonds.

Application

16:0 Diether PC or 1,2-di-O-hexadecyl-sn-glycero-3-phosphocholine might be used:
  • to study the miscibility and phase behavior of the cationic 1,2-dipalmitoyl-sn-glycero-3-ethylphosphocholine (EDPPC)
  • in unilamellar vesicles to study glycolipid transfer protein (GLTP)-mediated transfer of anthrylvinyl-galactosylceramide (AV-GalCer) from different ether phospholipid matrices
  • as a non-ladderane phosphocholine (PC) lipid reference sample in reversed-phase liquid chromatography (LC) method to separate PC lipids
  • to prepare large phospholipid bicelles in order to determine the three-dimensional backbone structure of the transmembrane domain of Vpu from human immunodeficiency virus (HIV)-1 by solid-state nuclear magnetic resonance (NMR) spectroscopy

Biochem/physiol Actions

Ether glycerolipids serve as membrane molecules and cellular mediators. Ether linked phospholipids act as precursors for important signaling molecules. 16:0 Diether PC (phosphocholine) is known to interdigitate even in the absence of chemical inducers or pressure.

Packaging

5 mL Amber Glass Screw Cap Vial (999992P-10mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ether-linked lipids and their bioactive species
Biochimica et Biophysica Acta - Biomembranes, 9(2), 245-276 (2008)
Three-dimensional structure of the transmembrane domain of Vpu from HIV-1 in aligned phospholipid bicelles
Park SH, et al.
Biophysical Journal, 91(8), 3032-3042 (2006)
Analysis of intact ladderane phospholipids, originating from viable anammox bacteria, using RP-LC-ESI-MS
Lanekoff I and Karlsson R
Analytical and Bioanalytical Chemistry, 397(8), 3543-3551 (2010)
Molecular features of phospholipids that affect glycolipid transfer protein-mediated galactosylceramide transfer between vesicles
Nylund M, et al.
Biochimica et Biophysica Acta - Biomembranes, 1758(6), 807-812 (2006)
The interdigitated gel phase in mixtures of cationic and zwitterionic phospholipids
Smith EA and Dea PH
Biophysical Chemistry, 196(2), 86-91 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service