Skip to Content
Merck
All Photos(1)

Key Documents

700000P

Avanti

Cholesterol (ovine)

Avanti Research - A Croda Brand

Synonym(s):

cholest-5-en-3β-ol ; 110796

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H46O
CAS Number:
Molecular Weight:
386.65
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

description

cholesterol (ovine wool, 98%)

Assay

>98% (TLC)

form

powder

packaging

pkg of 1 × 1 g (700000P-1g)
pkg of 1 × 100 mg (700000P-100mg)
pkg of 1 × 5 g (700000P-5g)
pkg of 1 × 500 mg (700000P-500mg)

manufacturer/tradename

Avanti Research - A Croda Brand

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@H]1CC[C@@]2([C@@H]3[C@H]([C@H]4[C@@]([C@H](CC4)[C@@H](CCCC(C)C)C)(CC3)C)CC=C2C1)C

InChI

1S/C27H46O/c1-18(2)7-6-8-19(3)23-11-12-24-22-10-9-20-17-21(28)13-15-26(20,4)25(22)14-16-27(23,24)5/h9,18-19,21-25,28H,6-8,10-17H2,1-5H3/t19-,21+,22+,23-,24+,25+,26+,27-/m1/s1

InChI key

HVYWMOMLDIMFJA-DPAQBDIFSA-N

Packaging

125 mL Amber Wide Mouth Glass Bottle with Screw Cap (700000P-5g)
20 mL Clear Glass Screw Cap Vial (700000P-1g)
20 mL Clear Glass Screw Cap Vial (700000P-500mg)
5 mL Amber Glass Screw Cap Vial (700000P-100mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roy Ziblat et al.
Biophysical journal, 103(2), 255-264 (2012-08-03)
Grazing incidence x-ray diffraction measurements were performed on single hydrated bilayers and monolayers of Ceramide/Cholesterol/1-palmitoyl-2-oleoyl-sn-glycero-3-phosphocyholine at varying concentrations. There are substantial differences in the phase and structure behavior of the crystalline domains formed within the bilayers relative to the corresponding
Izabella Brand et al.
Langmuir : the ACS journal of surfaces and colloids, 34(46), 14022-14032 (2018-10-27)
Many cytoplasmic proteins contain a hydrophobic acyl chain, which facilitates protein binding to cell membranes. Hydrophobic interactions between the exposed acyl chain of the protein and hydrocarbon chains of lipids in the cell membrane are the driving force for this
Ieva Vasiliauskaité-Brooks et al.
Nature, 544(7648), 120-123 (2017-03-23)
Adiponectin receptors (ADIPORs) are integral membrane proteins that control glucose and lipid metabolism by mediating, at least in part, a cellular ceramidase activity that catalyses the hydrolysis of ceramide to produce sphingosine and a free fatty acid (FFA). The crystal
Lukas Frey et al.
Journal of the American Chemical Society, 140(45), 15402-15411 (2018-10-06)
Membrane protein function fundamentally depends on lipid-bilayer fluidity and the composition of the biological membrane. Although dynamic interdependencies between membrane proteins and the surrounding lipids are suspected, a detailed description is still missing. To uncover lipid-modulated membrane protein backbone dynamics
Valerija Vezočnik et al.
Langmuir : the ACS journal of surfaces and colloids, 34(30), 8983-8993 (2018-07-10)
Understanding of the interactions between proteins and natural and artificially prepared lipid membrane surfaces and embedded nonpolar cores is important in studies of physiological processes and their pathologies and is applicable to nanotechnologies. In particular, rapidly growing interest in cellular

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service