Skip to Content
Merck
All Photos(1)

Key Documents

880120C

Avanti

18:0 PEG2000 PE

Avanti Research - A Croda Brand 880120C

Synonym(s):

1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-2000] (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

form

liquid

packaging

pkg of 1 × 2.5 mL (880120C-25mg)
pkg of 1 × 20 mL (880120C-500mg)
pkg of 2 × 4 mL (880120C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 880120C

concentration

10 mg/mL (880120C-25mg)
25 mg/mL (880120C-200mg)
25 mg/mL (880120C-500mg)

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@@](COP([O-])(OCCNC(OCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOC)=O)=O)(OC(CCCCCCCCCCCCCCCCC)=O)COC(CCCCCCCCCCCCCCCCC)=O.[NH4+]

InChI

1S/C133H264NO55P.H3N/c1-4-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-131(135)186-128-130(189-132(136)35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-5-2)129-188-190(138,139)187-37-36-134-133(137)185-127-126-184-125-124-183-123-122-182-121-120-181-119-118-180-1

InChI key

QSWJINHFEMDVEM-OJCMNYADSA-N

General description

18:0 PEG2000 PE, also known as 1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-2000], is a modified phospholipid containing a 2000-Da PEG moiety in the polar head.[1]

Application

1,2-distearoyl-sn-glycero-3-phosphoethanolamine-N-[methoxy(polyethylene glycol)-2000] ammonium salt (DSPE-PEG1) has been used to conjugate PEG onto residual amine groups on the surfaces of the functionalized TaO nanoparticles.[2] DSPE-PEG1 may be used as a surfactant for homeotropic anchoring of the mesogen molecules.[1] It may also be used in preparation of Nano formulations.[3]

Biochem/physiol Actions

PEG-modified lipids are typically added to phospholipid vesicles used for drug delivery to facilitate the formation of sterically stabilized liposomes (SSL). These liposomes have low reticuloendothelial system (RES) uptake and they prolong the circulating plasma half-life of the drug. The plasma half-life of the drug can be tuned from several hours to days depending on the size of the PEG and fatty acids attached to the lipid anchor.[4]

Packaging

30 mL Amber Narrow Mouth Glass Bottle with Screw Cap (880120C-500mg)
5 mL Clear Glass Sealed Ampule (880120C-200mg)
5 mL Clear Glass Sealed Ampule (880120C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Encapsulated miR-200c and Nkx2. 1 in a nuclear/mitochondria transcriptional regulatory network of non-metastatic and metastatic lung cancer cells
D?Almeida O, et al.
BMC Cancer, 19(1), 136-136 (2019)
Liposomes with detachable polymer coating: destabilization and fusion of dioleoylphosphatidylethanolamine vesicles triggered by cleavage of surface-grafted poly (ethylene glycol)
Kirpotin D, et al.
Febs Letters, 388(2-3), 115-118 (1996)
Active nematic emulsions
Guillamat P, et al.
Science Advances, 4(4), eaao1470-eaao1470 (2018)
P A Melnikov et al.
Bulletin of experimental biology and medicine (2019-11-24)
We studied the effect of IL-1β on the permeability of brain capillaries in healthy mice. Intravital microscopy demonstrated that parenteral administration of IL-1β was followed by an increase in vascular permeability ensuring passage of free Alexa488 fluorescent label through the
Ling Ye et al.
Nanotheranostics, 3(1), 54-65 (2019-01-22)
Rationale: Despite growing use of engineered nanomaterials (ENM) in applications from electronics to medicine, the potential risk to human health remains a critical concern within clinical use. ENM exposure during pregnancy can potentially cause reproductive toxicity even at levels that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service