Skip to Content
Merck
All Photos(1)

Key Documents

860524P

Avanti

C24 Ceramide (d18:1/24:0)

Avanti Research - A Croda Brand

Synonym(s):

N-lignoceroyl-D-erythro-sphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H83NO3
CAS Number:
Molecular Weight:
650.11
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 10 mg (860524P-10mg)
pkg of 1 × 5 mg (860524P-5mg)
pkg of 1 × 50 mg (860524P-50mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

OC[C@]([H])(NC(CCCCCCCCCCCCCCCCCCCCCCC)=O)[C@]([H])(O)/C=C/CCCCCCCCCCCCC

InChI

1S/C42H83NO3/c1-3-5-7-9-11-13-15-17-18-19-20-21-22-23-24-26-28-30-32-34-36-38-42(46)43-40(39-44)41(45)37-35-33-31-29-27-25-16-14-12-10-8-6-4-2/h35,37,40-41,44-45H,3-34,36,38-39H2,1-2H3,(H,43,46)/b37-35+/t40-,41+/m0/s1

InChI key

ZJVVOYPTFQEGPH-AUTSUKAISA-N

General description

C24 Ceramide has long acyl chain length of 24 carbon units.

Application

C24 Ceramide (d18:1/24:0) has been used:
  • in the generation of small unilamellar vesicles
  • as an internal standard in liquid chromatography-mass spectrometry for ceramide quantification from rat samples
  • to test its effect on mitochondrial stress response in worms

Biochem/physiol Actions

C24 ceramide induces mitochondrial stress response. Its levels are depleted in coronary artery disease (CAD). However, elevated ceramide levels is observed in Alzheimer′s disease. C24 is synthesized during stress. It mediates spontaneous neutrophil death.

Packaging

5 mL Amber Glass Screw Cap Vial (860524P-10mg)
5 mL Amber Glass Screw Cap Vial (860524P-50mg)
5 mL Amber Glass Screw Cap Vial (860524P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Permeability and microstructure of model stratum corneum lipid membranes containing ceramides with long (C16) and very long (C24) acyl chains
Pullmannova P, et al.
Biophysical Chemistry, 224, 20-31 (2017)
Ceramides and sphingomyelinases in senile plaques
Panchal M, et al.
Neurobiology of Disease, 65, 193-201 (2014)
Liposome Preparation for the Analysis of Lipid-Receptor Interaction and Efferocytosis
Voss OH, et al.
Current Protocols in Immunology, 120(1), 14-44 (2018)
Lukáš Opálka et al.
Journal of lipid research, 61(2), 219-228 (2019-12-21)
Ceramides (Cers) with ultralong (∼32-carbon) chains and ω-esterified linoleic acid, composing a subclass called omega-O-acylceramides (acylCers), are indispensable components of the skin barrier. Normal barriers typically contain acylCer concentrations of ∼10 mol%; diminished concentrations, along with altered or missing long
Estradiol regulates energy balance by ameliorating hypothalamic ceramide-induced ER Stress
Gonzalez-Garcia I, et al.
Cell Reports, 25(2), 413-423 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service