Skip to Content
Merck
All Photos(2)

Documents

850467C

Avanti

18:0-18:1 PC

1-stearoyl-2-oleoyl-sn-glycero-3-phosphocholine, chloroform

Synonym(s):

1-octadecanoyl-2--(9Z-octadecenoyl)-sn-glycero-3-phosphocholine; SOPC; PC(18:0/18:1(9Z)).

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H86NO8P
CAS Number:
Molecular Weight:
788.13
UNSPSC Code:
51191904
NACRES:
NA.25

Assay

>99% (TLC)

form

liquid

packaging

pkg of 1 × 2.5 mL (850467C-25mg)
pkg of 2 × 4 mL (850467C-200mg)

manufacturer/tradename

Avanti Research - A Croda Brand 850467C

concentration

10 mg/mL (850467C-25mg)
25 mg/mL (850467C-200mg)

lipid type

cardiolipins
phospholipids

shipped in

dry ice

storage temp.

−20°C

InChI

1S/C44H86NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-43(46)50-40-42(41-52-54(48,49)51-39-38-45(3,4)5)53-44(47)37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h21,23,42H,6-20,22,24-41H2,1-5H3/b23-21-/t42-/m1/s1

InChI key

ATHVAWFAEPLPPQ-VRDBWYNSSA-N

General description

Phosphatidylcholine (PC) is an important constituent of the mucosal layer of the colon. 18:0-18:1 PC (1-stearoyl-2-oleoyl-sn-glycero-3-phosphocholine) has a phosphocholine backbone.

Application

18:0-18:1 PC (1-stearoyl-2-oleoyl-sn-glycero-3-phosphocholine) may be used:
  • in liposomes for liposome binding assays
  • to prepare giant unilamellar vesicles (GUVs) by electroformation
  • to prepare lipid vesicles

Biochem/physiol Actions

Phosphatidylcholine (PC) plays a major role in endocytosis and the catalytic activity of enzymes.

Packaging

5 mL Clear Glass Sealed Ampule (850467C-200mg)
5 mL Clear Glass Sealed Ampule (850467C-25mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Integrative Medicine - E-Book, 125016-125016 null
Journal of Physical Science, 253(1), 012058-012058 null
Experimental study of the electrically induced optical birefringence in suspension of densely packed giant lipid vesicles
Antonova K, et al.
Test null
Compact and tunable size-based dielectrophoretic flow fractionation
Chuang HS, et al.
Journal of Micromechanics and Microengineering, 24(12), 125016-125016 null
Experimental study of the electrically induced optical birefringence in suspension of densely packed giant lipid vesicles
Antonova K, et al.
The Journal of Biological Chemistry null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service