Skip to Content
Merck
All Photos(3)

Key Documents

840523P

Avanti

MGDG

Avanti Research - A Croda Brand

Synonym(s):

1,2-diacyl-3-O-β-D-galactosyl-sn-glycerol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H74O10
CAS Number:
Molecular Weight:
775.06
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25
Pricing and availability is not currently available.

description

Monogalactosyldiacylglycerol (Plant)

Assay

>99% (TLC)

form

powder

packaging

pkg of 1 × 5 mg (840523P-5mg)
pkg of 2 × 5 mg (840523P-10mg)
pkg of 5 × 5 mg (840523P-25mg)

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

phosphoglycerides
bioactive lipids

shipped in

dry ice

storage temp.

−20°C

General description

Monogalactosyldiacylglycerol (MGDG) and digalactosyldiacylglycerol (DGDG) are the two nonionic lipid constituents of the thylakoid membrane of higher plant chloroplasts. MGDG and DGDG are present in the membrane at 56% and 29%, respectively, of the total lipid content. DGDG is a bilayer-forming lipid, while MGDG alone will only form hexagonal-II structures.
Monogalactosyldiacylglycerol (MGDG) synthesis involves generation of monoglucosyldiacylglycerol (MGlcDG) from diacylglycerol (DAG) in the presence of MGlcDG synthase. The second step is epimerization of MGlcDG to MGDG.[1] It forms nonbilayer structures[2] and is also associated with algae and fungi.[3]

Application

Monogalactosyldiacylglycerol (MGDG) may be used:
  • in bicelles preparation for nuclear magnetic resonance studies[2]
  • in the generation of structured MGDG for pinolenic acid enrichment[3]
  • in the preparation of liposomes[4][5]
  • as a lipid standard in hydrophilic interaction liquid chromatography-ion mobility mass spectrometry (HILIC-IM-MS)[6]

Biochem/physiol Actions

Monogalactosyldiacylglycerol (MGDG) synthesis is essential for chloroplast biogenesis. It is also needed for synchronizing photosynthesis associated plastid and nucleus related gene expression.[7] MGDG is essential for membrane lipid homeostasis during stress.[2] In food industry, MGDG serves as an enhancer in baking and as an non-ionic emulsifier.[3]

Packaging

5 mL Amber Glass Screw Cap Vial (840523P-10mg)
5 mL Amber Glass Screw Cap Vial (840523P-25mg)
5 mL Amber Glass Screw Cap Vial (840523P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

also commonly purchased with this product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Monogalactosyldiacylglycerol facilitates synthesis of photoactive protochlorophyllide in etioplasts
Fujii S, et al.
Plant Physiology, 174(4), 2183-2198 (2017)
Enzymatic Synthesis of Structured Monogalactosyldiacylglycerols Enriched in Pinolenic Acid
Kim J, et al.
Journal of Agricultural and Food Chemistry, 66(30), 8079-8085 (2018)
Fluorescence Microscopy of Single Liposomes with Incorporated Pigment-Proteins
Tutkus M, et al.
Langmuir, 34(47), 14410-14418 (2018)
Identification of plant-like galactolipids in Chromera velia, a photosynthetic relative of malaria parasites
BotteCY, et al.
The Journal of Biological Chemistry, 286(34), 29893-29903 (2011)
A Janus-faced IM30 ring involved in thylakoid membrane fusion is assembled from IM30 tetramers
Saur M, et al.
Structure, 25(9), 1380-1390 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service